Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574091_m1
          See other LRAT GE Assays ›
          Gene Symbol:
          Lrat
          Gene Name:
          lecithin-retinol acyltransferase (phosphatidylcholine-retinol-O-acyltransferase)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 181896304 - 181905366 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574091_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022280.2 NP_071616.1 1 - 2 805 91
          GenBank mRNA AF255060.2 - 1 - 2 805 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003938.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrat

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          vitamin A metabolic process
          visual perception
          positive regulation of lipid transport
          retinol metabolic process
          retinoic acid metabolic process
          retinoic acid binding
          O-acyltransferase activity
          retinol binding
          phosphatidylcholine-retinol O-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550748 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P491942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550748 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550748 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491942 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550748 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P491942 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491942 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28649 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823951 - -
          Affymetrix Rat Gene 1.0 ST Array 10823953 - -
          Affymetrix Rat Gene 1.0 ST Array 10823950 - -
          Affymetrix Rat Gene 1.0 ST Array 10823954 - -
          Affymetrix Rat Gene 1.0 ST Array 10823952 - -
          Affymetrix Rat Gene 1.1 ST Array 10823949 - -
          Rat Genome 230 2.0 Array 1368570_at - -
          Rat Genome 230 2.0 Array 1368570_PM_at - -
          Affymetrix HT Rat Focus Array 1368570_at - -
          Affymetrix HT Rat Focus Array 1368570_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200026468.rn.1 TC0200003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026467.rn.1 TC0200003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026466.rn.1 TC0200003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026469.rn.1 TC0200003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026464.rn.1 TC0200003938.rn.1 RTA TC
          Rat Expression Set 230 1368570_PM_at - -
          Rat Expression Set 230 1368570_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823949 - -
          Rat Genome U34 Set rc_AA899076_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22003184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline