Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574703_m1
          See other GPX3 GE Assays ›
          Gene Symbol:
          Gpx3
          Gene Name:
          glutathione peroxidase 3
          Gene Aliases:
          GPx-3, GPx-P, GSHPx-3, GSHPx-P, Gpxp
          Chromosome Location:
          Chr.10: 40247436 - 40255423 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022525.3 NP_071970.2 1 - 2 156 86
          GenBank mRNA BC062227.1 - 1 - 2 132 6921653 86
          CK480346.1 - 1 - 2 156 86
          D00680.1 - 1 - 2 221 86
          FQ221037.1 - 1 - 2 128 86
          FQ221279.1 - 1 - 2 109 86
          FQ228485.1 - 1 - 2 128 86
          FQ228545.1 - 1 - 2 182 86
          FQ228679.1 - 1 - 2 128 86
          FQ228689.1 - 1 - 2 128 86
          FQ228690.1 - 1 - 2 141 86
          FQ228694.1 - 1 - 2 108 86
          FQ228697.1 - 1 - 2 128 86
          FQ228705.1 - 1 - 2 126 86
          FQ228712.1 - 1 - 2 128 86
          FQ228719.1 - 1 - 2 128 86
          FQ228751.1 - 1 - 2 128 86
          FQ228788.1 - 1 - 2 128 86
          FQ228802.1 - 1 - 2 128 86
          FQ228804.1 - 1 - 2 128 86
          FQ228807.1 - 1 - 2 129 86
          FQ228808.1 - 1 - 2 128 86
          FQ228824.1 - 1 - 2 128 86
          FQ228834.1 - 1 - 2 130 86
          FQ228835.1 - 1 - 2 128 86
          FQ228845.1 - 1 - 2 128 86
          FQ228866.1 - 1 - 2 128 86
          FQ228878.1 - 1 - 2 128 86
          FQ228879.1 - 1 - 2 128 86
          FQ228886.1 - 1 - 2 128 86
          FQ228899.1 - 1 - 2 128 86
          FQ228911.1 - 1 - 2 86
          FQ228914.1 - 1 - 2 128 86
          FQ228921.1 - 1 - 2 128 86
          FQ228934.1 - 1 - 2 130 86
          FQ228945.1 - 1 - 2 128 86
          FQ228947.1 - 1 - 2 128 86
          FQ228960.1 - 1 - 2 128 86
          FQ228965.1 - 1 - 2 128 86
          FQ228969.1 - 1 - 2 127 86
          FQ228970.1 - 1 - 2 128 86
          FQ228976.1 - 1 - 2 128 86
          FQ229118.1 - 1 - 2 128 86
          FQ229125.1 - 1 - 2 128 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000614.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpx3

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to molecule of fungal origin
          glutathione metabolic process
          response to oxidative stress
          female pregnancy
          response to organic substance
          response to selenium ion
          response to organic cyclic compound
          response to drug
          hydrogen peroxide catabolic process
          protein homotetramerization
          response to corticosterone
          oxidation-reduction process
          glutathione peroxidase activity
          selenium binding
          glutathione binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline