Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575360_g1
          See other SCT GE Assays ›
          Gene Symbol:
          Sct
          Gene Name:
          secretin
          Gene Aliases:
          Secr
          Chromosome Location:
          Chr.1: 214264865 - 214270337 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575360_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sct
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022670.2 NP_073161.1 2 - 3 189 105
          XM_006230518.2 XP_006230580.1 3 - 4 437 105
          GenBank mRNA M31495.1 - 2 - 3 212 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006942.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sct

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          visual learning
          dentate gyrus development
          positive regulation of cAMP biosynthetic process
          negative regulation of neuron apoptotic process
          embryonic digestive tract development
          positive regulation of pancreatic juice secretion
          positive regulation of somatostatin secretion
          neuronal stem cell population maintenance
          negative regulation of gastrin-induced gastric acid secretion
          receptor binding
          hormone activity
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15427 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15427 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15427 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15427 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15427 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726790 - -
          Affymetrix Rat Gene 1.0 ST Array 10726793 - -
          Affymetrix Rat Gene 1.0 ST Array 10726791 - -
          Affymetrix Rat Gene 1.0 ST Array 10726792 - -
          Affymetrix Rat Gene 1.0 ST Array 10726789 - -
          Affymetrix Rat Gene 1.1 ST Array 10726788 - -
          Rat Genome 230 2.0 Array 1387257_at - -
          Rat Genome 230 2.0 Array 1387257_PM_at - -
          Affymetrix HT Rat Focus Array 1387257_at - -
          Affymetrix HT Rat Focus Array 1387257_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100054751.rn.1 TC0100006942.rn.1 RTA TC
          Rat Expression Set 230 1387257_at - -
          Rat Expression Set 230 1387257_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726788 - -
          Rat Genome U34 Set M64033_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21307205 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372135 - -
          RatRef-12 Expression BeadChip ILMN_63518 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline