Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575570_m1
          See other OLAH GE Assays ›
          Gene Symbol:
          Olah
          Gene Name:
          oleoyl-ACP hydrolase
          Gene Aliases:
          Mch, Thedc1
          Chromosome Location:
          Chr.17: 78878817 - 78903919 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575570_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olah
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022705.1 NP_073196.1 4 - 5 614 80
          XM_008771886.1 XP_008770108.1 5 - 6 768 80
          XM_008771887.1 XP_008770109.1 4 - 5 471 80
          GenBank mRNA M16200.1 - 4 - 5 614 80
          Y00311.1 - 3 - 4 349 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001029.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olah

          Panther Classification:

          Molecular Function -

          esterase hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          oleoyl-[acyl-carrier-protein] hydrolase activity
          myristoyl-[acyl-carrier-protein] hydrolase activity
          palmitoyl-[acyl-carrier-protein] hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299571 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95881 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796426 - -
          Affymetrix Rat Gene 1.0 ST Array 10796421 - -
          Affymetrix Rat Gene 1.0 ST Array 10796423 - -
          Affymetrix Rat Gene 1.0 ST Array 10796420 - -
          Affymetrix Rat Gene 1.0 ST Array 10796422 - -
          Affymetrix Rat Gene 1.0 ST Array 10796425 - -
          Affymetrix Rat Gene 1.0 ST Array 10796424 - -
          Affymetrix Rat Gene 1.0 ST Array 10796419 - -
          Affymetrix Rat Gene 1.0 ST Array 10796427 - -
          Affymetrix Rat Gene 1.1 ST Array 10796418 - -
          Rat Genome 230 2.0 Array 1368756_PM_at - -
          Rat Genome 230 2.0 Array 1368756_at - -
          Affymetrix HT Rat Focus Array 1368756_at - -
          Affymetrix HT Rat Focus Array 1368756_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700007075.rn.1 TC1700001029.rn.1 RTA TC
          Rat Expression Set 230 1368756_at - -
          Rat Expression Set 230 1368756_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796418 - -
          Rat Genome U34 Set Y00311_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22241918 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371132 - -
          RatRef-12 Expression BeadChip ILMN_70174 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline