Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576640_m1
          See other RAB7A GE Assays ›
          Gene Symbol:
          Rab7a
          Gene Name:
          RAB7A, member RAS oncogene family
          Gene Aliases:
          Rab7
          Chromosome Location:
          Chr.4: 119910466 - 119963065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576640_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023950.3 NP_076440.1 2 - 3 217 71
          XM_006236849.2 XP_006236911.1 2 - 3 168 71
          GenBank mRNA AB158427.1 - 1 - 2 49 71
          AB158428.1 - 1 - 2 49 71
          AF286535.1 - 2 - 3 213 71
          BC072470.1 - 2 - 3 217 7098657 71
          X12535.1 - 2 - 3 90 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003282.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab7a

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to lysosome
          intracellular protein transport
          epidermal growth factor catabolic process
          endosome to lysosome transport
          protein to membrane docking
          Rab protein signal transduction
          retrograde transport, endosome to Golgi
          early endosome to late endosome transport
          bone resorption
          phagosome acidification
          phagosome-lysosome fusion
          positive regulation of exosomal secretion
          regulation of autophagosome assembly
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1053383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053383 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053383 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1053383 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1053383 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863879 - -
          Affymetrix Rat Gene 1.0 ST Array 10863878 - -
          Affymetrix Rat Gene 1.0 ST Array 10863880 - -
          Affymetrix Rat Gene 1.0 ST Array 10863883 - -
          Affymetrix Rat Gene 1.0 ST Array 10863882 - -
          Affymetrix Rat Gene 1.0 ST Array 10863881 - -
          Affymetrix Rat Gene 1.0 ST Array 10863877 - -
          Affymetrix Rat Gene 1.1 ST Array 10863876 - -
          Rat Genome 230 2.0 Array 1387797_PM_at - -
          Rat Genome 230 2.0 Array 1387797_at - -
          Rat Genome 230 2.0 Array 1398838_PM_at - -
          Rat Genome 230 2.0 Array 1398838_at - -
          Affymetrix HT Rat Focus Array 1398838_at - -
          Affymetrix HT Rat Focus Array 1398838_PM_at - -
          Affymetrix HT Rat Focus Array 1387797_at - -
          Affymetrix HT Rat Focus Array 1387797_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400023373.rn.1 TC0400003282.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023369.rn.1 TC0400003282.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023368.rn.1 TC0400003282.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023370.rn.1 TC0400003282.rn.1 RTA TC
          Rat Expression Set 230 1387797_PM_at - -
          Rat Expression Set 230 1398838_PM_at - -
          Rat Expression Set 230 1387797_at - -
          Rat Expression Set 230 1398838_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863876 - -
          Rat Genome U34 Set X12535cds_at - -
          Rat Genome U34 Set rc_AA946057_at - -
          Rat Genome U34 Set rc_AA818669_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21587648 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374078 - -
          RatRef-12 Expression BeadChip ILMN_66929 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline