Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577100_m1
          See other HAP1 GE Assays ›
          Gene Symbol:
          Hap1
          Gene Name:
          huntingtin-associated protein 1
          Gene Aliases:
          HAP1-A, HAP1-B
          Chromosome Location:
          Chr.10: 88257975 - 88266210 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024133.2 NP_077047.1 6 - 7 958 65
          NM_177982.1 NP_817091.1 6 - 7 958 65
          GenBank mRNA U38370.1 - 6 - 7 973 65
          U38373.1 - 6 - 7 973 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003791.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hap1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          autophagy
          anterograde axonal transport
          retrograde axonal transport
          protein localization
          positive regulation of neuron projection development
          protein transport
          regulation of exocytosis
          cerebellum development
          hypothalamus cell differentiation
          cell projection organization
          neuron projection development
          positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          positive regulation of synaptic transmission, GABAergic
          positive regulation of neurotrophin production
          early endosome to late endosome transport
          positive regulation of epidermal growth factor receptor signaling pathway
          vesicle transport along microtubule
          neurotrophin TRK receptor signaling pathway
          positive regulation of neurogenesis
          cellular response to growth factor stimulus
          positive regulation of inclusion body assembly
          negative regulation of beta-amyloid formation
          regulation of organelle transport along microtubule
          positive regulation of nonmotile primary cilium assembly
          receptor binding
          protein binding
          protein domain specific binding
          identical protein binding
          ion channel binding
          brain-derived neurotrophic factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P111723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241811 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25703 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P111723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P111723 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P111723 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P111723 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700511495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12592 - -
          Agilent Rat cDNA Microarray (G4105A) 700511495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747328 - -
          Affymetrix Rat Gene 1.0 ST Array 10747326 - -
          Affymetrix Rat Gene 1.0 ST Array 10747318 - -
          Affymetrix Rat Gene 1.0 ST Array 10747327 - -
          Affymetrix Rat Gene 1.0 ST Array 10747321 - -
          Affymetrix Rat Gene 1.0 ST Array 10747329 - -
          Affymetrix Rat Gene 1.0 ST Array 10747315 - -
          Affymetrix Rat Gene 1.0 ST Array 10747317 - -
          Affymetrix Rat Gene 1.0 ST Array 10747322 - -
          Affymetrix Rat Gene 1.0 ST Array 10747320 - -
          Affymetrix Rat Gene 1.0 ST Array 10747316 - -
          Affymetrix Rat Gene 1.0 ST Array 10747324 - -
          Affymetrix Rat Gene 1.0 ST Array 10747325 - -
          Affymetrix Rat Gene 1.0 ST Array 10747314 - -
          Affymetrix Rat Gene 1.0 ST Array 10747323 - -
          Affymetrix Rat Gene 1.1 ST Array 10747313 - -
          Rat Genome 230 2.0 Array 1369358_PM_a_at - -
          Rat Genome 230 2.0 Array 1369358_a_at - -
          Affymetrix HT Rat Focus Array 1369358_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369358_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000033908.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033914.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033900.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033897.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033899.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033912.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033901.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033905.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033907.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033902.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033903.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033904.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033910.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033913.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033896.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033911.rn.1 TC1000003791.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000033915.rn.1 TC1000003791.rn.1 RTA TC
          Rat Expression Set 230 1369358_PM_a_at - -
          Rat Expression Set 230 1369358_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747313 - -
          Rat Genome U34 Set U38373_s_at - -
          Rat Genome U34 Set U38373_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20760796 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375614 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376870 - -
          RatRef-12 Expression BeadChip ILMN_66361 - -
          RatRef-12 Expression BeadChip ILMN_70064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline