Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577377_m1
          See other AK1 GE Assays ›
          Gene Symbol:
          Ak1
          Gene Name:
          adenylate kinase 1
          Gene Aliases:
          Ak 1
          Chromosome Location:
          Chr.3: 11652143 - 11659135 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577377_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024349.3 NP_077325.2 367 102
          GenBank mRNA AB022701.1 - 329 102
          BC089797.1 - 367 7130672 102
          D13376.1 - 117 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000264.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ak1

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleoside diphosphate phosphorylation
          ADP biosynthetic process
          cell cycle arrest
          muscle organ development
          nucleoside triphosphate biosynthetic process
          positive regulation of glucose transport
          positive regulation of neuron maturation
          response to activity
          cerebellum development
          olfactory bulb development
          neuron differentiation
          response to estradiol
          response to testosterone
          response to drug
          AMP metabolic process
          ATP metabolic process
          adenine metabolic process
          inosine biosynthetic process
          nucleotide phosphorylation
          adenylate kinase activity
          nucleoside diphosphate kinase activity
          ATP binding
          phosphotransferase activity, phosphate group as acceptor

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P835462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P835462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P835462 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P835462 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12416 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701745390 - -
          Agilent Rat cDNA Microarray (G4105A) 701745390 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940629 - -
          Affymetrix Rat Gene 1.1 ST Array 10940629 - -
          Rat Genome 230 2.0 Array 1370011_PM_at - -
          Rat Genome 230 2.0 Array 1389185_at - -
          Rat Genome 230 2.0 Array 1389185_PM_at - -
          Rat Genome 230 2.0 Array 1370011_at - -
          Affymetrix HT Rat Focus Array 1389185_at - -
          Affymetrix HT Rat Focus Array 1389185_PM_at - -
          Affymetrix HT Rat Focus Array 1370011_PM_at - -
          Affymetrix HT Rat Focus Array 1370011_at - -
          Affymetrix Rat Clariom S Assay PSR0300003001.rn.1 TC0300000264.rn.1 RTA TC
          Rat Expression Set 230 1389185_PM_at - -
          Rat Expression Set 230 1370011_PM_at - -
          Rat Expression Set 230 1370011_at - -
          Rat Expression Set 230 1389185_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940629 - -
          Rat Genome U34 Set rc_AA799299_at - -
          Rat Genome U34 Set D13376_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21499277 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline