Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578534_m1
          See other SNAP25 GE Assays ›
          Gene Symbol:
          Snap25
          Gene Name:
          synaptosomal-associated protein 25
          Gene Aliases:
          SNAP-25B, SNAP-25a
          Chromosome Location:
          Chr.3: 129697408 - 129788417 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578534_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snap25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270575.1 NP_001257504.1 5 - 6 621 104
          NM_001270576.1 NP_001257505.1 5 - 6 533 104
          NM_030991.3 NP_112253.1 5 - 6 621 104
          GenBank mRNA AB003992.1 - 4 - 5 412 104
          AF245227.1 - 5 - 6 620 104
          BC087699.1 - 5 - 6 576 7314318 104
          DY315301.1 - 5 - 6 553 104
          FM033263.1 - 5 - 6 533 104
          FQ212040.1 - 5 - 6 592 104
          FQ213289.1 - 6 - 7 104
          FQ213923.1 - 5 - 6 592 104
          FQ214162.1 - 5 - 6 621 104
          U56261.1 - 385 104
          U56262.1 - 4 - 5 385 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001811.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snap25

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          neurotransmitter secretion
          axonogenesis
          long-term memory
          regulation of neuron projection development
          synaptic vesicle priming
          endosomal transport
          growth hormone secretion
          sleep
          synaptic vesicle fusion to presynaptic active zone membrane
          positive regulation of insulin secretion
          positive regulation of hormone secretion
          calcium ion-regulated exocytosis of neurotransmitter
          regulation of synapse assembly
          long-term synaptic potentiation
          regulation of establishment of protein localization
          potassium ion transmembrane transport
          SNARE binding
          voltage-gated potassium channel activity
          SNAP receptor activity
          protein binding
          myosin binding
          syntaxin-1 binding
          protein domain specific binding
          protein N-terminus binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019966 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P371172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P371172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P371172 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019966 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P371172 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P371172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48100 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840250 - -
          Affymetrix Rat Gene 1.0 ST Array 10840253 - -
          Affymetrix Rat Gene 1.0 ST Array 10840251 - -
          Affymetrix Rat Gene 1.0 ST Array 10840254 - -
          Affymetrix Rat Gene 1.0 ST Array 10840252 - -
          Affymetrix Rat Gene 1.0 ST Array 10840246 - -
          Affymetrix Rat Gene 1.0 ST Array 10840247 - -
          Affymetrix Rat Gene 1.0 ST Array 10840248 - -
          Affymetrix Rat Gene 1.1 ST Array 10840245 - -
          Rat Genome 230 2.0 Array 1387073_PM_at - -
          Rat Genome 230 2.0 Array 1387073_at - -
          Affymetrix HT Rat Focus Array 1387073_at - -
          Affymetrix HT Rat Focus Array 1387073_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300013118.rn.1 TC0300001811.rn.1 RTA TC
          Rat Expression Set 230 1387073_PM_at - -
          Rat Expression Set 230 1387073_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840245 - -
          Rat Genome U34 Set AB003992_s_at - -
          Rat Genome U34 Set AB003991_at - -
          Rat Genome U34 Set U56261_s_at - -
          Rat Genome U34 Set AB003991_g_at - -
          Rat Genome U34 Set rc_AA943153_at - -
          Rat Genome U34 Set AB003992_s_at - -
          Rat Genome U34 Set AB003991_g_at - -
          Rat Genome U34 Set U56261_s_at - -
          Rat Genome U34 Set AB003991_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22421320 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348840 - -
          RatRef-12 Expression BeadChip ILMN_52755 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline