Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578948_m1
          See other GAK GE Assays ›
          Gene Symbol:
          Gak
          Gene Name:
          cyclin G associated kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 2100091 - 2174332 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578948_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gak
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031030.2 NP_112292.1 7 - 8 844 68
          XM_006250583.2 XP_006250645.1 7 - 8 859 68
          XM_006250584.2 XP_006250646.1 7 - 8 859 68
          GenBank mRNA D38560.1 - 7 - 8 845 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000033.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gak

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          endoplasmic reticulum organization
          Golgi organization
          epidermal cell differentiation
          intrahepatic bile duct development
          cell development
          forebrain morphogenesis
          establishment of skin barrier
          clathrin coat disassembly
          clathrin-mediated endocytosis
          positive regulation of neural precursor cell proliferation
          protein serine/threonine kinase activity
          ATP binding
          cyclin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14928 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14928 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14928 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51822 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770953 - -
          Affymetrix Rat Gene 1.0 ST Array 10770954 - -
          Affymetrix Rat Gene 1.0 ST Array 10770965 - -
          Affymetrix Rat Gene 1.0 ST Array 10770970 - -
          Affymetrix Rat Gene 1.0 ST Array 10770975 - -
          Affymetrix Rat Gene 1.0 ST Array 10770964 - -
          Affymetrix Rat Gene 1.0 ST Array 10770957 - -
          Affymetrix Rat Gene 1.0 ST Array 10770960 - -
          Affymetrix Rat Gene 1.0 ST Array 10763427 - -
          Affymetrix Rat Gene 1.0 ST Array 10770974 - -
          Affymetrix Rat Gene 1.0 ST Array 10770956 - -
          Affymetrix Rat Gene 1.0 ST Array 10770959 - -
          Affymetrix Rat Gene 1.0 ST Array 10770963 - -
          Affymetrix Rat Gene 1.0 ST Array 10770973 - -
          Affymetrix Rat Gene 1.0 ST Array 10770967 - -
          Affymetrix Rat Gene 1.0 ST Array 10770949 - -
          Affymetrix Rat Gene 1.0 ST Array 10770961 - -
          Affymetrix Rat Gene 1.0 ST Array 10770969 - -
          Affymetrix Rat Gene 1.0 ST Array 10770951 - -
          Affymetrix Rat Gene 1.0 ST Array 10770972 - -
          Affymetrix Rat Gene 1.0 ST Array 10770952 - -
          Affymetrix Rat Gene 1.0 ST Array 10770950 - -
          Affymetrix Rat Gene 1.0 ST Array 10770962 - -
          Affymetrix Rat Gene 1.0 ST Array 10770971 - -
          Affymetrix Rat Gene 1.0 ST Array 10770968 - -
          Affymetrix Rat Gene 1.0 ST Array 10770955 - -
          Affymetrix Rat Gene 1.0 ST Array 10770966 - -
          Affymetrix Rat Gene 1.1 ST Array 10770948 - -
          Affymetrix Rat Gene 1.1 ST Array 10763426 - -
          Rat Genome 230 2.0 Array 1367875_at - -
          Rat Genome 230 2.0 Array 1367875_PM_at - -
          Affymetrix HT Rat Focus Array 1367875_at - -
          Affymetrix HT Rat Focus Array 1367875_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400000256.rn.1 TC1400000033.rn.1 RTA TC
          Rat Expression Set 230 1367875_PM_at - -
          Rat Expression Set 230 1367875_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770948 - -
          Rat Genome U34 Set D38560_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336073 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356475 - -
          RatRef-12 Expression BeadChip ILMN_48602 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline