Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579211_m1
          See other MUSK GE Assays ›
          Gene Symbol:
          Musk
          Gene Name:
          muscle, skeletal, receptor tyrosine kinase
          Gene Aliases:
          Nsk1
          Chromosome Location:
          Chr.5: 75392790 - 75498694 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Musk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031061.1 NP_112323.1 4 - 5 605 102
          GenBank mRNA U34985.1 - 4 - 5 605 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000716.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Musk

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          regulation of transcription, DNA-templated
          neuromuscular junction development
          memory
          regulation of synaptic growth at neuromuscular junction
          positive regulation of gene expression
          negative regulation of gene expression
          response to muscle activity
          peptidyl-tyrosine phosphorylation
          cell differentiation
          positive regulation of synaptic transmission, cholinergic
          receptor clustering
          positive regulation of neuron apoptotic process
          positive regulation of synaptic growth at neuromuscular junction
          protein autophosphorylation
          response to axon injury
          positive regulation of peptidyl-tyrosine phosphorylation
          response to electrical stimulus
          retina development in camera-type eye
          long-term synaptic potentiation
          skeletal muscle acetylcholine-gated channel clustering
          cochlea development
          positive regulation of protein geranylgeranylation
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126719 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12498 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12498 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10869227 - -
          Affymetrix Rat Gene 1.0 ST Array 10869228 - -
          Affymetrix Rat Gene 1.0 ST Array 10869225 - -
          Affymetrix Rat Gene 1.0 ST Array 10869223 - -
          Affymetrix Rat Gene 1.0 ST Array 10869222 - -
          Affymetrix Rat Gene 1.0 ST Array 10869214 - -
          Affymetrix Rat Gene 1.0 ST Array 10869221 - -
          Affymetrix Rat Gene 1.0 ST Array 10869215 - -
          Affymetrix Rat Gene 1.0 ST Array 10869224 - -
          Affymetrix Rat Gene 1.0 ST Array 10869229 - -
          Affymetrix Rat Gene 1.0 ST Array 10869226 - -
          Affymetrix Rat Gene 1.0 ST Array 10869218 - -
          Affymetrix Rat Gene 1.0 ST Array 10869219 - -
          Affymetrix Rat Gene 1.0 ST Array 10869216 - -
          Affymetrix Rat Gene 1.0 ST Array 10869217 - -
          Affymetrix Rat Gene 1.0 ST Array 10869230 - -
          Affymetrix Rat Gene 1.1 ST Array 10869213 - -
          Rat Genome 230 2.0 Array 1369388_at - -
          Rat Genome 230 2.0 Array 1369388_PM_at - -
          Affymetrix HT Rat Focus Array 1369388_PM_at - -
          Affymetrix HT Rat Focus Array 1369388_at - -
          Affymetrix Rat Clariom S Assay PSR0500004783.rn.1 TC0500000716.rn.1 RTA TC
          Rat Expression Set 230 1369388_at - -
          Rat Expression Set 230 1369388_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10869213 - -
          Rat Genome U34 Set U34985_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21351516 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359905 - -
          RatRef-12 Expression BeadChip ILMN_54862 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline