Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579752_m1
          See other DOC2B GE Assays ›
          Gene Symbol:
          Doc2b
          Gene Name:
          double C2-like domains, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 63924969 - 63952948 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579752_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Doc2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031142.1 NP_112404.1 3 - 4 685 81
          XM_008768117.1 XP_008766339.1 3 - 4 746 81
          GenBank mRNA U70778.2 - 3 - 4 685 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003288.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Doc2b

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein localization
          positive regulation of vesicle fusion
          positive regulation of insulin secretion
          positive regulation of calcium ion-dependent exocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          transporter activity
          calcium-dependent phospholipid binding
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P131749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132512 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P131749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P131749 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P131749 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P131749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744976 - -
          Affymetrix Rat Gene 1.0 ST Array 10744978 - -
          Affymetrix Rat Gene 1.0 ST Array 10744977 - -
          Affymetrix Rat Gene 1.0 ST Array 10744973 - -
          Affymetrix Rat Gene 1.0 ST Array 10744975 - -
          Affymetrix Rat Gene 1.0 ST Array 10744974 - -
          Affymetrix Rat Gene 1.0 ST Array 10744979 - -
          Affymetrix Rat Gene 1.0 ST Array 10744971 - -
          Affymetrix Rat Gene 1.0 ST Array 10744972 - -
          Affymetrix Rat Gene 1.0 ST Array 10744980 - -
          Affymetrix Rat Gene 1.1 ST Array 10744970 - -
          Rat Genome 230 2.0 Array 1369479_at - -
          Rat Genome 230 2.0 Array 1369479_PM_at - -
          Affymetrix HT Rat Focus Array 1369479_at - -
          Affymetrix HT Rat Focus Array 1369479_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000029374.rn.1 TC1000003288.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029375.rn.1 TC1000003288.rn.1 RTA TC
          Rat Expression Set 230 1369479_at - -
          Rat Expression Set 230 1369479_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744970 - -
          Rat Genome U34 Set U70778_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21792475 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364148 - -
          RatRef-12 Expression BeadChip ILMN_66841 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline