Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580055_m1
          See other CYR61 GE Assays ›
          Gene Symbol:
          Cyr61
          Gene Name:
          cysteine-rich, angiogenic inducer, 61
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 251529354 - 251532312 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580055_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyr61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031327.2 NP_112617.2 1 - 2 248 113
          GenBank mRNA AB015877.1 - 1 - 2 113 113
          AF218568.1 - 1 - 2 248 113
          BC081689.1 - 1 - 2 228 7122117 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004857.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyr61

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          osteoblast differentiation
          positive regulation of protein phosphorylation
          intussusceptive angiogenesis
          atrioventricular valve morphogenesis
          apoptotic process involved in heart morphogenesis
          ventricular septum development
          chemotaxis
          cell adhesion
          signal transduction
          cell-cell signaling
          positive regulation of phospholipase activity
          positive regulation of cell-substrate adhesion
          single organismal cell-cell adhesion
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          positive regulation of osteoblast proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          wound healing, spreading of cells
          positive regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          positive regulation of protein kinase activity
          positive regulation of transcription from RNA polymerase II promoter
          atrial septum morphogenesis
          negative regulation of cell death
          chondroblast differentiation
          chorio-allantoic fusion
          labyrinthine layer blood vessel development
          positive regulation of cartilage development
          regulation of ERK1 and ERK2 cascade
          reactive oxygen species metabolic process
          positive regulation of ceramide biosynthetic process
          integrin binding
          insulin-like growth factor binding
          heparin binding
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0200033625.rn.1 TC0200004857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033624.rn.1 TC0200004857.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline