Search
Search
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031344.2 | NP_112634.1 | 7 - 8 | 1050 | 100 | |
| XM_008760244.1 | XP_008758466.1 | 7 - 8 | 2559 | 100 | ||
| GenBank mRNA | AB021980.1 | - | 7 - 8 | 980 | 100 | |
| BC081776.1 | - | 7 - 8 | 1050 | 7128630 | 100 | |
| FQ210893.1 | - | 7 - 8 | 979 | 100 | ||
| HQ909027.1 | - | 7 - 8 | 884 | 100 | ||
Set Membership: |
|