Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580900_m1
          See other IGHMBP2 GE Assays ›
          Gene Symbol:
          Ighmbp2
          Gene Name:
          immunoglobulin mu binding protein 2
          Gene Aliases:
          AEP
          Chromosome Location:
          Chr.1: 218509274 - 218531922 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ighmbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031586.1 NP_113774.1 13 - 14 2593 92
          GenBank mRNA AF199411.1 - 13 - 14 2593 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007030.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ighmbp2

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          metabolic process
          spinal cord motor neuron differentiation
          DNA duplex unwinding
          protein homooligomerization
          tRNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          RNA binding
          ATP binding
          DNA-dependent ATPase activity
          transcription factor binding
          RNA-dependent ATPase activity
          zinc ion binding
          ATP-dependent 5'-3' RNA helicase activity
          ribosome binding
          ATP-dependent 5'-3' DNA helicase activity
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072035 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072035 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072035 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P559041 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52905 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727312 - -
          Affymetrix Rat Gene 1.0 ST Array 10727310 - -
          Affymetrix Rat Gene 1.0 ST Array 10727304 - -
          Affymetrix Rat Gene 1.0 ST Array 10727311 - -
          Affymetrix Rat Gene 1.0 ST Array 10727314 - -
          Affymetrix Rat Gene 1.0 ST Array 10727305 - -
          Affymetrix Rat Gene 1.0 ST Array 10727308 - -
          Affymetrix Rat Gene 1.0 ST Array 10727316 - -
          Affymetrix Rat Gene 1.0 ST Array 10727307 - -
          Affymetrix Rat Gene 1.0 ST Array 10727309 - -
          Affymetrix Rat Gene 1.0 ST Array 10727313 - -
          Affymetrix Rat Gene 1.0 ST Array 10727306 - -
          Affymetrix Rat Gene 1.0 ST Array 10727319 - -
          Affymetrix Rat Gene 1.0 ST Array 10727318 - -
          Affymetrix Rat Gene 1.0 ST Array 10727320 - -
          Affymetrix Rat Gene 1.0 ST Array 10727315 - -
          Affymetrix Rat Gene 1.0 ST Array 10727317 - -
          Affymetrix Rat Gene 1.1 ST Array 10727303 - -
          Rat Genome 230 2.0 Array 1368855_PM_at - -
          Rat Genome 230 2.0 Array 1368855_at - -
          Affymetrix HT Rat Focus Array 1368855_at - -
          Affymetrix HT Rat Focus Array 1368855_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100055772.rn.1 TC0100007030.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055773.rn.1 TC0100007030.rn.1 RTA TC
          Rat Expression Set 230 1368855_at - -
          Rat Expression Set 230 1368855_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727303 - -
          Rat Genome U34 Set rc_AI170531_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21190153 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374282 - -
          RatRef-12 Expression BeadChip ILMN_69832 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline