Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580949_m1
          See other P2RX4 GE Assays ›
          Gene Symbol:
          P2rx4
          Gene Name:
          purinergic receptor P2X, ligand-gated ion channel 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 39308022 - 39325673 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580949_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2rx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031594.1 NP_113782.1 1 - 2 231 67
          GenBank mRNA BC078792.1 - 1 - 2 172 7123247 67
          U32497.1 - 1 - 2 208 67
          U47031.1 - 1 - 2 161 67
          X87763.1 - 1 - 2 231 67
          X91200.1 - 1 - 2 146 67
          X93565.1 - 1 - 2 318 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001585.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2rx4

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of sodium ion transport
          nitric oxide biosynthetic process
          transport
          calcium ion transport
          signal transduction
          regulation of blood pressure
          positive regulation of calcium ion transport into cytosol
          negative regulation of cardiac muscle hypertrophy
          neuronal action potential
          sensory perception of pain
          response to ATP
          ion transmembrane transport
          response to fluid shear stress
          purinergic nucleotide receptor signaling pathway
          vasodilation
          positive regulation of calcium-mediated signaling
          protein homooligomerization
          membrane depolarization
          regulation of cardiac muscle contraction
          relaxation of cardiac muscle
          excitatory postsynaptic potential
          cellular response to ATP
          apoptotic signaling pathway
          cation transmembrane transport
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          receptor binding
          copper ion binding
          protein binding
          ATP binding
          drug binding
          zinc ion binding
          cadherin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P484909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110406 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P484909 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484909 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700929177 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700273828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98356 - -
          Agilent Rat cDNA Microarray (G4105A) 700929177 - -
          Agilent Rat cDNA Microarray (G4105A) 700273828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761981 - -
          Affymetrix Rat Gene 1.0 ST Array 10761982 - -
          Affymetrix Rat Gene 1.0 ST Array 10761991 - -
          Affymetrix Rat Gene 1.0 ST Array 10761983 - -
          Affymetrix Rat Gene 1.0 ST Array 10761980 - -
          Affymetrix Rat Gene 1.0 ST Array 10761985 - -
          Affymetrix Rat Gene 1.0 ST Array 10761988 - -
          Affymetrix Rat Gene 1.0 ST Array 10761989 - -
          Affymetrix Rat Gene 1.0 ST Array 10761990 - -
          Affymetrix Rat Gene 1.0 ST Array 10761987 - -
          Affymetrix Rat Gene 1.0 ST Array 10761979 - -
          Affymetrix Rat Gene 1.0 ST Array 10761984 - -
          Affymetrix Rat Gene 1.0 ST Array 10761986 - -
          Affymetrix Rat Gene 1.1 ST Array 10761978 - -
          Rat Genome 230 2.0 Array 1369743_PM_a_at - -
          Rat Genome 230 2.0 Array 1369743_a_at - -
          Affymetrix HT Rat Focus Array 1369743_a_at - -
          Affymetrix HT Rat Focus Array 1369743_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1200013501.rn.1 TC1200001585.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013503.rn.1 TC1200001585.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013504.rn.1 TC1200001585.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200013506.rn.1 TC1200001585.rn.1 RTA TC
          Rat Expression Set 230 1369743_PM_a_at - -
          Rat Expression Set 230 1369743_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761978 - -
          Rat Genome U34 Set U47031_at - -
          Rat Genome U34 Set U47031_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20832048 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366191 - -
          RatRef-12 Expression BeadChip ILMN_68777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline