Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581276_m1
          See other HPGDS GE Assays ›
          Gene Symbol:
          Hpgds
          Gene Name:
          hematopoietic prostaglandin D synthase
          Gene Aliases:
          Ptgds2
          Chromosome Location:
          Chr.4: 95945512 - 95970666 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpgds
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031644.2 NP_113832.1 4 - 5 692 80
          XM_006236602.2 XP_006236664.1 4 - 5 660 80
          XM_006236603.2 XP_006236665.1 3 - 4 582 80
          GenBank mRNA BC087590.1 - 4 - 5 692 7307804 80
          D82071.1 - 4 - 5 523 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003016.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpgds

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          prostaglandin metabolic process
          negative regulation of male germ cell proliferation
          magnesium ion binding
          glutathione transferase activity
          prostaglandin-D synthase activity
          calcium ion binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P439292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162053 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P439292 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P439292 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P439292 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P439292 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701436231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91901 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46073 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81938 - -
          Agilent Rat cDNA Microarray (G4105A) 701436231 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862845 - -
          Affymetrix Rat Gene 1.0 ST Array 10862847 - -
          Affymetrix Rat Gene 1.0 ST Array 10862844 - -
          Affymetrix Rat Gene 1.0 ST Array 10862849 - -
          Affymetrix Rat Gene 1.0 ST Array 10862843 - -
          Affymetrix Rat Gene 1.0 ST Array 10862848 - -
          Affymetrix Rat Gene 1.0 ST Array 10862846 - -
          Affymetrix Rat Gene 1.1 ST Array 10862842 - -
          Rat Genome 230 2.0 Array 1368492_PM_at - -
          Rat Genome 230 2.0 Array 1368492_at - -
          Affymetrix HT Rat Focus Array 1368492_PM_at - -
          Affymetrix HT Rat Focus Array 1368492_at - -
          Affymetrix Rat Clariom S Assay PSR0400021380.rn.1 TC0400003016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021382.rn.1 TC0400003016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021379.rn.1 TC0400003016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021383.rn.1 TC0400003016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021386.rn.1 TC0400003016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021384.rn.1 TC0400003016.rn.1 RTA TC
          Rat Expression Set 230 1368492_PM_at - -
          Rat Expression Set 230 1368492_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862842 - -
          Rat Genome U34 Set D82071_at - -
          Rat Genome U34 Set AF021882_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22276997 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371397 - -
          RatRef-12 Expression BeadChip ILMN_63349 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline