Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582112_m1
          See other ALCAM GE Assays ›
          Gene Symbol:
          Alcam
          Gene Name:
          activated leukocyte cell adhesion molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 50781127 - 50985083 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582112_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alcam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031753.1 NP_113941.1 1 - 2 256 78
          GenBank mRNA AB008538.1 - 1 - 2 256 78
          BC061970.1 - 1 - 2 265 5621856 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000511.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alcam

          Gene Ontology Categories:

          Function(s)

          cell adhesion
          axon guidance
          motor neuron axon guidance

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P135277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47929 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135277 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P135277 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12662 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135277 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700812848 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78557 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17303 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54540 - -
          Agilent Rat cDNA Microarray (G4105A) 700812848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750883 - -
          Affymetrix Rat Gene 1.0 ST Array 10750884 - -
          Affymetrix Rat Gene 1.0 ST Array 10750892 - -
          Affymetrix Rat Gene 1.0 ST Array 10750894 - -
          Affymetrix Rat Gene 1.0 ST Array 10750882 - -
          Affymetrix Rat Gene 1.0 ST Array 10750891 - -
          Affymetrix Rat Gene 1.0 ST Array 10750887 - -
          Affymetrix Rat Gene 1.0 ST Array 10750888 - -
          Affymetrix Rat Gene 1.0 ST Array 10750889 - -
          Affymetrix Rat Gene 1.0 ST Array 10750893 - -
          Affymetrix Rat Gene 1.0 ST Array 10750881 - -
          Affymetrix Rat Gene 1.0 ST Array 10750895 - -
          Affymetrix Rat Gene 1.0 ST Array 10750885 - -
          Affymetrix Rat Gene 1.0 ST Array 10750886 - -
          Affymetrix Rat Gene 1.0 ST Array 10750880 - -
          Affymetrix Rat Gene 1.0 ST Array 10750879 - -
          Affymetrix Rat Gene 1.0 ST Array 10750890 - -
          Affymetrix Rat Gene 1.1 ST Array 10750878 - -
          Rat Genome 230 2.0 Array 1370043_PM_at - -
          Rat Genome 230 2.0 Array 1370043_at - -
          Rat Genome 230 2.0 Array 1374432_PM_at - -
          Rat Genome 230 2.0 Array 1374432_at - -
          Rat Genome 230 2.0 Array 1382294_PM_at - -
          Rat Genome 230 2.0 Array 1382294_at - -
          Affymetrix HT Rat Focus Array 1370043_at - -
          Affymetrix HT Rat Focus Array 1370043_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100002606.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002618.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002604.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002615.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002616.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002607.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002609.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002605.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002608.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002617.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002622.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002623.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002628.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002621.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002614.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002619.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002625.rn.1 TC1100000511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002624.rn.1 TC1100000511.rn.1 RTA TC
          Rat Expression Set 230 1374432_at - -
          Rat Expression Set 230 1370043_PM_at - -
          Rat Expression Set 230 1374432_PM_at - -
          Rat Expression Set 230 1370043_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750878 - -
          Rat Genome U34 Set AB008538_at - -
          Rat Genome U34 Set X95850mRNA_g_at - -
          Rat Genome U34 Set X95850mRNA_at - -
          Rat Genome U34 Set rc_AI137483_at - -
          Rat Genome U34 Set rc_AI170117_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21099979 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370154 - -
          RatRef-12 Expression BeadChip ILMN_67898 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline