Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582505_m1
          See other SLC12A2 GE Assays ›
          Gene Symbol:
          Slc12a2
          Gene Name:
          solute carrier family 12 (sodium/potassium/chloride transporter), member 2
          Gene Aliases:
          Bsc2, Nkcc1
          Chromosome Location:
          Chr.18: 52917124 - 52985281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc12a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031798.1 NP_113986.1 17 - 18 2717 67
          XM_006254752.2 XP_006254814.1 17 - 18 2648 67
          GenBank mRNA AF051561.1 - 17 - 18 2717 67
          AF071863.1 - 17 - 18 2000 67
          FQ211948.1 - 1 - 2 81 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000572.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc12a2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          sodium ion transport
          chloride transport
          hyperosmotic response
          gamma-aminobutyric acid signaling pathway
          aging
          ammonium transport
          transepithelial chloride transport
          multicellular organism growth
          positive regulation of cell volume
          detection of mechanical stimulus involved in sensory perception of sound
          branching involved in mammary gland duct morphogenesis
          mammary duct terminal end bud growth
          transepithelial ammonium transport
          ammonium transmembrane transport
          chloride transmembrane transport
          sodium:potassium:chloride symporter activity
          ammonium transmembrane transporter activity
          cation:chloride symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P163595 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P742882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163595 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P742882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P742882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163595 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P742882 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P163595 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P134405 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163408 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700522202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103004 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88455 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22465 - -
          Agilent Rat cDNA Microarray (G4105A) 700522202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801897 - -
          Affymetrix Rat Gene 1.0 ST Array 10801903 - -
          Affymetrix Rat Gene 1.0 ST Array 10801888 - -
          Affymetrix Rat Gene 1.0 ST Array 10801886 - -
          Affymetrix Rat Gene 1.0 ST Array 10801889 - -
          Affymetrix Rat Gene 1.0 ST Array 10801910 - -
          Affymetrix Rat Gene 1.0 ST Array 10801909 - -
          Affymetrix Rat Gene 1.0 ST Array 10801898 - -
          Affymetrix Rat Gene 1.0 ST Array 10801901 - -
          Affymetrix Rat Gene 1.0 ST Array 10801900 - -
          Affymetrix Rat Gene 1.0 ST Array 10801887 - -
          Affymetrix Rat Gene 1.0 ST Array 10801907 - -
          Affymetrix Rat Gene 1.0 ST Array 10801908 - -
          Affymetrix Rat Gene 1.0 ST Array 10801902 - -
          Affymetrix Rat Gene 1.0 ST Array 10801885 - -
          Affymetrix Rat Gene 1.0 ST Array 10801895 - -
          Affymetrix Rat Gene 1.0 ST Array 10801911 - -
          Affymetrix Rat Gene 1.0 ST Array 10801896 - -
          Affymetrix Rat Gene 1.0 ST Array 10801899 - -
          Affymetrix Rat Gene 1.0 ST Array 10801890 - -
          Affymetrix Rat Gene 1.0 ST Array 10801905 - -
          Affymetrix Rat Gene 1.0 ST Array 10801913 - -
          Affymetrix Rat Gene 1.0 ST Array 10801894 - -
          Affymetrix Rat Gene 1.0 ST Array 10801912 - -
          Affymetrix Rat Gene 1.0 ST Array 10801914 - -
          Affymetrix Rat Gene 1.0 ST Array 10801891 - -
          Affymetrix Rat Gene 1.0 ST Array 10801892 - -
          Affymetrix Rat Gene 1.0 ST Array 10801906 - -
          Affymetrix Rat Gene 1.0 ST Array 10801904 - -
          Affymetrix Rat Gene 1.0 ST Array 10801893 - -
          Affymetrix Rat Gene 1.1 ST Array 10801884 - -
          Rat Genome 230 2.0 Array 1367853_PM_at - -
          Rat Genome 230 2.0 Array 1367853_at - -
          Affymetrix HT Rat Focus Array 1367853_PM_at - -
          Affymetrix HT Rat Focus Array 1367853_at - -
          Affymetrix Rat Clariom S Assay PSR1800004201.rn.1 TC1800000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004202.rn.1 TC1800000572.rn.1 RTA TC
          Rat Expression Set 230 1367853_PM_at - -
          Rat Expression Set 230 1367853_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801884 - -
          Rat Genome U34 Set S82233_s_at - -
          Rat Genome U34 Set AF086758_at - -
          Rat Genome U34 Set AF051561_s_at - -
          Rat Genome U34 Set AF086758_g_at - -
          Rat Genome U34 Set rc_AA925961_s_at - -
          Rat Genome U34 Set rc_AI072983_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21130848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline