Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583189_m1
          See other IMPA1 GE Assays ›
          Gene Symbol:
          Impa1
          Gene Name:
          Inositol (myo)-1(or 4)-monophosphatase 1
          Gene Aliases:
          Impa1-L
          Chromosome Location:
          Chr.2: 93675203 - 93696355 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583189_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Impa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032057.1 NP_114446.1 4 - 5 378 59
          XM_006232151.2 XP_006232213.1 4 - 5 537 59
          GenBank mRNA U84038.2 - 4 - 5 378 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000775.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Impa1

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol biosynthetic process
          phosphatidylinositol biosynthetic process
          phosphate-containing compound metabolic process
          signal transduction
          response to lithium ion
          phosphatidylinositol phosphorylation
          inositol phosphate dephosphorylation
          magnesium ion binding
          inositol monophosphate 1-phosphatase activity
          manganese ion binding
          lithium ion binding
          identical protein binding
          protein homodimerization activity
          inositol monophosphate 3-phosphatase activity
          inositol monophosphate 4-phosphatase activity
          inositol monophosphate phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12712 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12712 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12712 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12712 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P346750 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700506710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48832 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37888 - -
          Agilent Rat cDNA Microarray (G4105A) 700506710 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814277 - -
          Affymetrix Rat Gene 1.0 ST Array 10814278 - -
          Affymetrix Rat Gene 1.0 ST Array 10814276 - -
          Affymetrix Rat Gene 1.0 ST Array 10814271 - -
          Affymetrix Rat Gene 1.0 ST Array 10814273 - -
          Affymetrix Rat Gene 1.0 ST Array 10814274 - -
          Affymetrix Rat Gene 1.0 ST Array 10814275 - -
          Affymetrix Rat Gene 1.0 ST Array 10814272 - -
          Affymetrix Rat Gene 1.0 ST Array 10814280 - -
          Affymetrix Rat Gene 1.1 ST Array 10814270 - -
          Rat Genome 230 2.0 Array 1398808_at - -
          Rat Genome 230 2.0 Array 1398808_PM_at - -
          Affymetrix HT Rat Focus Array 1398808_PM_at - -
          Affymetrix HT Rat Focus Array 1398808_at - -
          Affymetrix Rat Clariom S Assay PSR0200004752.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004755.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004757.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004759.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004761.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004764.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004765.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004762.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004766.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004751.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004756.rn.1 TC0200000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004760.rn.1 TC0200000775.rn.1 RTA TC
          Rat Expression Set 230 1398808_at - -
          Rat Expression Set 230 1398808_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10814270 - -
          Rat Genome U34 Set rc_AA801286_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22354460 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364531 - -
          RatRef-12 Expression BeadChip ILMN_61486 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline