Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583517_m1
          See other LASP1 GE Assays ›
          Gene Symbol:
          Lasp1
          Gene Name:
          LIM and SH3 protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 85744662 - 85785130 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583517_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lasp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032613.2 NP_116002.1 6 - 7 705 82
          XM_006247288.2 XP_006247350.1 6 - 7 637 82
          GenBank mRNA AF242187.1 - 6 - 7 619 82
          BC099791.1 - 6 - 7 705 7934892 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lasp1

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          ion transmembrane transport
          zinc ion binding
          ion transmembrane transporter activity
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P162991 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P380381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107402 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P380381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P162991 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039444 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380381 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13548 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10737912 - -
          Affymetrix Rat Gene 1.0 ST Array 10737910 - -
          Affymetrix Rat Gene 1.0 ST Array 10737909 - -
          Affymetrix Rat Gene 1.0 ST Array 10737916 - -
          Affymetrix Rat Gene 1.0 ST Array 10737911 - -
          Affymetrix Rat Gene 1.0 ST Array 10737913 - -
          Affymetrix Rat Gene 1.0 ST Array 10737914 - -
          Affymetrix Rat Gene 1.0 ST Array 10737915 - -
          Affymetrix Rat Gene 1.1 ST Array 10737908 - -
          Rat Genome 230 2.0 Array 1388566_at - -
          Rat Genome 230 2.0 Array 1387844_at - -
          Rat Genome 230 2.0 Array 1388566_PM_at - -
          Rat Genome 230 2.0 Array 1387844_PM_at - -
          Affymetrix HT Rat Focus Array 1388566_at - -
          Affymetrix HT Rat Focus Array 1388566_PM_at - -
          Affymetrix HT Rat Focus Array 1387844_PM_at - -
          Affymetrix HT Rat Focus Array 1387844_at - -
          Affymetrix Rat Clariom S Assay PSR1000013787.rn.1 TC1000001555.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000013786.rn.1 TC1000001555.rn.1 RTA TC
          Rat Expression Set 230 1388566_at - -
          Rat Expression Set 230 1387844_PM_at - -
          Rat Expression Set 230 1387844_at - -
          Rat Expression Set 230 1388566_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10737908 - -
          Rat Genome U34 Set rc_AA901365_at - -
          Rat Genome U34 Set rc_AI013437_at - -
          Rat Genome U34 Set rc_AA901365_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21255746 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355228 - -
          RatRef-12 Expression BeadChip ILMN_62278 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline