Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584069_m1
          See other HPX GE Assays ›
          Gene Symbol:
          Hpx
          Gene Name:
          hemopexin
          Gene Aliases:
          Hpxn
          Chromosome Location:
          Chr.1: 170423558 - 170431066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584069_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053318.1 NP_445770.1 7 - 8 850 68
          GenBank mRNA BC091137.1 - 7 - 8 841 7326169 68
          FQ218998.1 - 4 - 5 619 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006294.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpx

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of immunoglobulin production
          positive regulation of humoral immune response mediated by circulating immunoglobulin
          transport
          hemoglobin metabolic process
          heme metabolic process
          positive regulation of tyrosine phosphorylation of Stat1 protein
          regulation of protein metabolic process
          positive regulation of response to interferon-gamma
          positive regulation of interferon-gamma-mediated signaling pathway
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93041 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037806 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1037806 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71213 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724484 - -
          Affymetrix Rat Gene 1.0 ST Array 10724485 - -
          Affymetrix Rat Gene 1.0 ST Array 10724493 - -
          Affymetrix Rat Gene 1.0 ST Array 10724488 - -
          Affymetrix Rat Gene 1.0 ST Array 10724490 - -
          Affymetrix Rat Gene 1.0 ST Array 10724489 - -
          Affymetrix Rat Gene 1.0 ST Array 10724486 - -
          Affymetrix Rat Gene 1.0 ST Array 10724487 - -
          Affymetrix Rat Gene 1.0 ST Array 10724492 - -
          Affymetrix Rat Gene 1.0 ST Array 10724491 - -
          Affymetrix Rat Gene 1.1 ST Array 10724483 - -
          Rat Genome 230 2.0 Array 1370065_PM_at - -
          Rat Genome 230 2.0 Array 1370065_at - -
          Affymetrix HT Rat Focus Array 1370065_PM_at - -
          Affymetrix HT Rat Focus Array 1370065_at - -
          Affymetrix Rat Clariom S Assay PSR0100049621.rn.1 TC0100006294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049622.rn.1 TC0100006294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049624.rn.1 TC0100006294.rn.1 RTA TC
          Rat Expression Set 230 1370065_PM_at - -
          Rat Expression Set 230 1370065_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724483 - -
          Rat Genome U34 Set M62642_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21257153 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363859 - -
          RatRef-12 Expression BeadChip ILMN_55688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline