Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00584298_m1
See other NR5A1 GE Assays ›
Gene Symbol:
Nr5a1
Gene Name:
nuclear receptor subfamily 5, group A, member 1
Gene Aliases:
Ad4BP/SF-1, Ad4bp, Ftzf1, Sf-1
Chromosome Location:
Chr.3: 22998900 - 23020441 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00584298_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Nr5a1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001191099.1
NP_001178028.1
4 - 5
1023
65
XM_006234106.2
XP_006234168.1
2 - 3
2034
65
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300003010.rn.1
Set Membership:
Amplicon length less than or equal to 70
Nuclear Receptor
3' Most
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
Gene Ontology Categories:
Function(s)
Process(es)
luteinization
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
male gonad development
female gonad development
tissue development
multicellular organism aging
reproductive process
cell differentiation
adrenal gland development
intracellular receptor signaling pathway
hormone metabolic process
steroid hormone mediated signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
maintenance of protein location in nucleus
positive regulation of male gonad development
negative regulation of female gonad development
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
phospholipid binding
zinc ion binding
enzyme binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P009788
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P009788
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P009788
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_43_P15478
-
-
Array Name
Probe IDs
Targets
Target Type
Rat Genome 230 2.0 Array
1393286_PM_at
-
-
Rat Genome 230 2.0 Array
1393286_at
-
-
Rat Genome 230 2.0 Array
1369859_PM_at
-
-
Rat Genome 230 2.0 Array
1369859_at
-
-
Affymetrix HT Rat Focus Array
1369859_at
-
-
Affymetrix HT Rat Focus Array
1369859_PM_at
-
-
Affymetrix Rat Clariom S Assay
PSR0300023340.rn.1
TC0300003010.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0300023338.rn.1
TC0300003010.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0300023339.rn.1
TC0300003010.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0300023341.rn.1
TC0300003010.rn.1
RTA TC
Rat Expression Set 230
1369859_PM_at
-
-
Rat Expression Set 230
1369859_at
-
-
Rat Expression Set 230
1393286_at
-
-
Rat Expression Set 230
1393286_PM_at
-
-
Rat Genome U34 Set
D42156cds_at
-
-
Rat Genome U34 Set
rc_AA850212_at
-
-
Rat Genome U34 Set
D42156cds_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21285620
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1649975
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline