Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584341_m1
          See other AGPS GE Assays ›
          Gene Symbol:
          Agps
          Gene Name:
          alkylglycerone phosphate synthase
          Gene Aliases:
          Adap-s, Adps, Ads
          Chromosome Location:
          Chr.3: 62648352 - 62749250 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584341_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Agps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053350.2 NP_445802.2 12 - 13 1532 98
          GenBank mRNA AF121052.1 - 12 - 13 1437 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000889.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Agps

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid biosynthetic process
          ether lipid biosynthetic process
          oxidation-reduction process
          alkylglycerone-phosphate synthase activity
          UDP-N-acetylmuramate dehydrogenase activity
          flavin adenine dinucleotide binding
          FAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112759 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112749 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132080 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93506 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12780 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12780 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112759 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12780 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112749 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112759 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 486 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837052 - -
          Affymetrix Rat Gene 1.0 ST Array 10837051 - -
          Affymetrix Rat Gene 1.1 ST Array 10837050 - -
          Rat Genome 230 2.0 Array 1370137_at - -
          Rat Genome 230 2.0 Array 1370137_PM_at - -
          Affymetrix HT Rat Focus Array 1370137_at - -
          Affymetrix HT Rat Focus Array 1370137_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300006612.rn.1 TC0300000889.rn.1 RTA TC
          Rat Expression Set 230 1370137_PM_at - -
          Rat Expression Set 230 1370137_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837050 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22283873 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline