Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584525_g1
          See other CD93 GE Assays ›
          Gene Symbol:
          Cd93
          Gene Name:
          CD93 molecule
          Gene Aliases:
          C1qRp, C1qr1, Ly68
          Chromosome Location:
          Chr.3: 142778798 - 142783774 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584525_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053383.1 NP_445835.1 1 - 2 2099 104
          GenBank mRNA AF136537.1 - 1 - 2 2026 104
          AF160978.1 - 1 - 2 2099 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004380.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd93

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          complement component C1q binding
          calcium ion binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12791 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12791 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12791 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850532 - -
          Affymetrix Rat Gene 1.0 ST Array 10850536 - -
          Affymetrix Rat Gene 1.0 ST Array 10850531 - -
          Affymetrix Rat Gene 1.0 ST Array 10850535 - -
          Affymetrix Rat Gene 1.0 ST Array 10850534 - -
          Affymetrix Rat Gene 1.0 ST Array 10850537 - -
          Affymetrix Rat Gene 1.0 ST Array 10850533 - -
          Affymetrix Rat Gene 1.0 ST Array 10850538 - -
          Affymetrix Rat Gene 1.1 ST Array 10850530 - -
          Rat Genome 230 2.0 Array 1387168_at - -
          Rat Genome 230 2.0 Array 1387168_PM_at - -
          Rat Genome 230 2.0 Array 1376574_at - -
          Rat Genome 230 2.0 Array 1376574_PM_at - -
          Rat Genome 230 2.0 Array 1368393_PM_at - -
          Rat Genome 230 2.0 Array 1368393_at - -
          Affymetrix HT Rat Focus Array 1368393_at - -
          Affymetrix HT Rat Focus Array 1387168_at - -
          Affymetrix HT Rat Focus Array 1368393_PM_at - -
          Affymetrix HT Rat Focus Array 1387168_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300033707.rn.1 TC0300004380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300033712.rn.1 TC0300004380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300033708.rn.1 TC0300004380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300033709.rn.1 TC0300004380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300033711.rn.1 TC0300004380.rn.1 RTA TC
          Rat Expression Set 230 1368393_PM_at - -
          Rat Expression Set 230 1368393_at - -
          Rat Expression Set 230 1387168_at - -
          Rat Expression Set 230 1387168_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10850530 - -
          Rat Genome U34 Set rc_AI011243_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20816044 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348939 - -
          RatRef-12 Expression BeadChip ILMN_58355 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline