Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584808_m1
          See other QSOX1 GE Assays ›
          Gene Symbol:
          Qsox1
          Gene Name:
          quiescin Q6 sulfhydryl oxidase 1
          Gene Aliases:
          Qscn6, Qsox, Sox-2, rSOx
          Chromosome Location:
          Chr.13: 73423396 - 73460890 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584808_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Qsox1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109898.1 NP_001103368.1 10 - 11 1324 75
          NM_053431.3 NP_445883.1 10 - 11 1324 75
          GenBank mRNA AB044285.1 - 10 - 11 1324 75
          AF217799.1 - 10 - 11 1328 75
          AF285078.1 - 10 - 11 1346 75
          AY623665.1 - 10 - 11 1326 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001685.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Qsox1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          cell redox homeostasis
          oxidation-reduction process
          protein disulfide isomerase activity
          flavin-linked sulfhydryl oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P258855 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P258855 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P258855 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093477 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P258855 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P258855 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768863 - -
          Affymetrix Rat Gene 1.0 ST Array 10768866 - -
          Affymetrix Rat Gene 1.0 ST Array 10768869 - -
          Affymetrix Rat Gene 1.0 ST Array 10768872 - -
          Affymetrix Rat Gene 1.0 ST Array 10768860 - -
          Affymetrix Rat Gene 1.0 ST Array 10768873 - -
          Affymetrix Rat Gene 1.0 ST Array 10768868 - -
          Affymetrix Rat Gene 1.0 ST Array 10768867 - -
          Affymetrix Rat Gene 1.0 ST Array 10768858 - -
          Affymetrix Rat Gene 1.0 ST Array 10768859 - -
          Affymetrix Rat Gene 1.0 ST Array 10768870 - -
          Affymetrix Rat Gene 1.0 ST Array 10768864 - -
          Affymetrix Rat Gene 1.0 ST Array 10768862 - -
          Affymetrix Rat Gene 1.0 ST Array 10768871 - -
          Affymetrix Rat Gene 1.0 ST Array 10768865 - -
          Affymetrix Rat Gene 1.0 ST Array 10768861 - -
          Affymetrix Rat Gene 1.1 ST Array 10768857 - -
          Rat Genome 230 2.0 Array 1368023_at - -
          Rat Genome 230 2.0 Array 1368024_PM_at - -
          Rat Genome 230 2.0 Array 1368024_at - -
          Rat Genome 230 2.0 Array 1368023_PM_at - -
          Affymetrix HT Rat Focus Array 1368023_PM_at - -
          Affymetrix HT Rat Focus Array 1368023_at - -
          Affymetrix HT Rat Focus Array 1368024_PM_at - -
          Affymetrix HT Rat Focus Array 1368024_at - -
          Affymetrix Rat Clariom S Assay PSR1300011644.rn.1 TC1300001685.rn.1 RTA TC
          Rat Expression Set 230 1368023_PM_at - -
          Rat Expression Set 230 1368023_at - -
          Rat Expression Set 230 1368024_PM_at - -
          Rat Expression Set 230 1368024_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768857 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21575196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360522 - -
          RatRef-12 Expression BeadChip ILMN_61482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline