Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584945_m1
          See other RAB27B GE Assays ›
          Gene Symbol:
          Rab27b
          Gene Name:
          RAB27B, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 68484927 - 68644585 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab27b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053459.1 NP_445911.1 4 - 5 576 79
          XM_006254930.2 XP_006254992.1 5 - 6 538 79
          GenBank mRNA AF325693.1 - 4 - 5 576 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001665.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab27b

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle docking involved in exocytosis
          metabolic process
          protein secretion
          regulation of exocytosis
          melanosome transport
          Rab protein signal transduction
          positive regulation of exocytosis
          multivesicular body sorting pathway
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          protein domain specific binding
          myosin V binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P114465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114465 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P114465 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P795294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P486055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76461 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26189 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805080 - -
          Affymetrix Rat Gene 1.0 ST Array 10805081 - -
          Affymetrix Rat Gene 1.0 ST Array 10805083 - -
          Affymetrix Rat Gene 1.0 ST Array 10805084 - -
          Affymetrix Rat Gene 1.0 ST Array 10805082 - -
          Affymetrix Rat Gene 1.0 ST Array 10805079 - -
          Affymetrix Rat Gene 1.1 ST Array 10805077 - -
          Rat Genome 230 2.0 Array 1370122_at - -
          Rat Genome 230 2.0 Array 1370122_PM_at - -
          Affymetrix HT Rat Focus Array 1370122_PM_at - -
          Affymetrix HT Rat Focus Array 1370122_at - -
          Affymetrix Rat Clariom S Assay PSR1800011698.rn.1 TC1800001665.rn.1 RTA TC
          Rat Expression Set 230 1370122_at - -
          Rat Expression Set 230 1370122_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805077 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21049118 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373113 - -
          RatRef-12 Expression BeadChip ILMN_57906 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline