Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585107_m1
          See other PIK3CB GE Assays ›
          Gene Symbol:
          Pik3cb
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 107275849 - 107363440 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585107_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3cb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006243642.2 XP_006243704.1 17 - 18 2521 63
          XM_008766567.1 XP_008764789.1 12 - 13 1908 63
          GenBank mRNA AJ012482.1 - 17 - 18 2504 63

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell-matrix adhesion
          cellular calcium ion homeostasis
          endocytosis
          autophagy
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          platelet activation
          phosphatidylinositol-3-phosphate biosynthetic process
          embryonic cleavage
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          phosphatidylinositol 3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81077 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P104679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41426 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919460 - -
          Affymetrix Rat Gene 1.0 ST Array 10919457 - -
          Affymetrix Rat Gene 1.0 ST Array 10919454 - -
          Affymetrix Rat Gene 1.0 ST Array 10919467 - -
          Affymetrix Rat Gene 1.0 ST Array 10820167 - -
          Affymetrix Rat Gene 1.0 ST Array 10820164 - -
          Affymetrix Rat Gene 1.0 ST Array 10919465 - -
          Affymetrix Rat Gene 1.0 ST Array 10919461 - -
          Affymetrix Rat Gene 1.0 ST Array 10820166 - -
          Affymetrix Rat Gene 1.0 ST Array 10919468 - -
          Affymetrix Rat Gene 1.0 ST Array 10820168 - -
          Affymetrix Rat Gene 1.0 ST Array 10919458 - -
          Affymetrix Rat Gene 1.0 ST Array 10919455 - -
          Affymetrix Rat Gene 1.0 ST Array 10919469 - -
          Affymetrix Rat Gene 1.0 ST Array 10919466 - -
          Affymetrix Rat Gene 1.0 ST Array 10820165 - -
          Affymetrix Rat Gene 1.0 ST Array 10919459 - -
          Affymetrix Rat Gene 1.0 ST Array 10919463 - -
          Affymetrix Rat Gene 1.0 ST Array 10919462 - -
          Affymetrix Rat Gene 1.0 ST Array 10919456 - -
          Affymetrix Rat Gene 1.1 ST Array 10820163 - -
          Affymetrix Rat Gene 1.1 ST Array 10919453 - -
          Rat Genome 230 2.0 Array 1387847_at - -
          Rat Genome 230 2.0 Array 1387847_PM_at - -
          Affymetrix HT Rat Focus Array 1387847_at - -
          Affymetrix HT Rat Focus Array 1387847_PM_at - -
          Rat Expression Set 230 1387847_at - -
          Rat Expression Set 230 1387847_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919453 - -
          Rat Genome U34 Set AJ012482_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20718184 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368644 - -
          RatRef-12 Expression BeadChip ILMN_66637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline