Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586096_m1
          See other BBS2 GE Assays ›
          Gene Symbol:
          Bbs2
          Gene Name:
          Bardet-Biedl syndrome 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 11385865 - 11421517 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586096_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bbs2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053618.1 NP_446070.1 12 - 13 1632 107
          XM_006255131.2 XP_006255193.1 12 - 13 1791 107
          XM_008772263.1 XP_008770485.1 12 - 13 1559 107
          GenBank mRNA AF342738.1 - 12 - 13 1632 107
          FQ222286.1 - 6 - 7 1063 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000108.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bbs2

          Gene Ontology Categories:

          Function(s) Process(es)

          sperm axoneme assembly
          protein localization
          biological_process
          negative regulation of gene expression
          artery smooth muscle contraction
          striatum development
          hippocampus development
          cerebral cortex development
          adult behavior
          leptin-mediated signaling pathway
          protein localization to organelle
          nonmotile primary cilium assembly
          negative regulation of appetite by leptin-mediated signaling pathway
          negative regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          vasodilation
          cilium assembly
          Golgi to plasma membrane protein transport
          response to leptin
          fat cell differentiation
          photoreceptor cell maintenance
          brain morphogenesis
          cartilage development
          cilium morphogenesis
          regulation of cilium beat frequency involved in ciliary motility
          protein localization to ciliary membrane
          RNA polymerase II repressing transcription factor binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12392 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12875 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12875 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12875 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33960 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806020 - -
          Affymetrix Rat Gene 1.0 ST Array 10806026 - -
          Affymetrix Rat Gene 1.0 ST Array 10806029 - -
          Affymetrix Rat Gene 1.0 ST Array 10806018 - -
          Affymetrix Rat Gene 1.0 ST Array 10806017 - -
          Affymetrix Rat Gene 1.0 ST Array 10806019 - -
          Affymetrix Rat Gene 1.0 ST Array 10806032 - -
          Affymetrix Rat Gene 1.0 ST Array 10806028 - -
          Affymetrix Rat Gene 1.0 ST Array 10806030 - -
          Affymetrix Rat Gene 1.0 ST Array 10806033 - -
          Affymetrix Rat Gene 1.0 ST Array 10806025 - -
          Affymetrix Rat Gene 1.0 ST Array 10806021 - -
          Affymetrix Rat Gene 1.0 ST Array 10806023 - -
          Affymetrix Rat Gene 1.0 ST Array 10806024 - -
          Affymetrix Rat Gene 1.0 ST Array 10806031 - -
          Affymetrix Rat Gene 1.0 ST Array 10806027 - -
          Affymetrix Rat Gene 1.0 ST Array 10806022 - -
          Affymetrix Rat Gene 1.1 ST Array 10806016 - -
          Rat Genome 230 2.0 Array 1368509_at - -
          Rat Genome 230 2.0 Array 1368509_PM_at - -
          Affymetrix HT Rat Focus Array 1368509_PM_at - -
          Affymetrix HT Rat Focus Array 1368509_at - -
          Affymetrix Rat Clariom S Assay PSR1900000919.rn.1 TC1900000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900000921.rn.1 TC1900000108.rn.1 RTA TC
          Rat Expression Set 230 1368509_at - -
          Rat Expression Set 230 1368509_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806016 - -
          Rat Genome U34 Set rc_AA945879_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22339395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372482 - -
          RatRef-12 Expression BeadChip ILMN_48331 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline