Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586375_m1
          See other IL5RA GE Assays ›
          Gene Symbol:
          Il5ra
          Gene Name:
          interleukin 5 receptor, alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 138798769 - 138830636 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586375_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il5ra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053645.1 NP_446097.1 3 - 4 355 92
          XM_006236977.2 XP_006237039.1 5 - 6 746 92
          GenBank mRNA AB056101.1 - 3 - 4 355 92
          AF324153.1 - 3 - 4 355 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003488.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il5ra

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response to antigenic stimulus
          cytokine-mediated signaling pathway
          regulation of interleukin-5 production
          cellular response to organic substance
          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143113 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046908 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046908 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046908 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16272 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864515 - -
          Affymetrix Rat Gene 1.0 ST Array 10864513 - -
          Affymetrix Rat Gene 1.0 ST Array 10864517 - -
          Affymetrix Rat Gene 1.0 ST Array 10864518 - -
          Affymetrix Rat Gene 1.0 ST Array 10864514 - -
          Affymetrix Rat Gene 1.0 ST Array 10864520 - -
          Affymetrix Rat Gene 1.0 ST Array 10864522 - -
          Affymetrix Rat Gene 1.0 ST Array 10864521 - -
          Affymetrix Rat Gene 1.0 ST Array 10864516 - -
          Affymetrix Rat Gene 1.0 ST Array 10864519 - -
          Affymetrix Rat Gene 1.1 ST Array 10864512 - -
          Rat Genome 230 2.0 Array 1387333_PM_at - -
          Rat Genome 230 2.0 Array 1387333_at - -
          Affymetrix HT Rat Focus Array 1387333_at - -
          Affymetrix HT Rat Focus Array 1387333_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400024549.rn.1 TC0400003488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024548.rn.1 TC0400003488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024547.rn.1 TC0400003488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024551.rn.1 TC0400003488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024550.rn.1 TC0400003488.rn.1 RTA TC
          Rat Expression Set 230 1387333_at - -
          Rat Expression Set 230 1387333_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864512 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22076244 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372052 - -
          RatRef-12 Expression BeadChip ILMN_69126 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline