Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586403_m1
          See other CXCL2 GE Assays ›
          Gene Symbol:
          Cxcl2
          Gene Name:
          chemokine (C-X-C motif) ligand 2
          Gene Aliases:
          Mip-2, Scyb2
          Chromosome Location:
          Chr.14: 18731346 - 18733391 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586403_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053647.1 NP_446099.1 3 - 4 354 85
          GenBank mRNA AB060092.1 - 3 - 4 354 85
          S77604.1 - 3 - 4 309 85
          U45965.1 - 3 - 4 317 85
          X65647.1 - 3 - 4 294 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001503.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl2

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          response to molecule of bacterial origin
          positive regulation of leukocyte chemotaxis
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          response to heat
          response to gamma radiation
          neutrophil chemotaxis
          leukocyte chemotaxis
          response to estradiol
          wound healing
          regulation of cell proliferation
          response to glucocorticoid
          chemokine-mediated signaling pathway
          neuron death
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          chemokine activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140437 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140437 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77918 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775898 - -
          Affymetrix Rat Gene 1.0 ST Array 10775899 - -
          Affymetrix Rat Gene 1.0 ST Array 10775897 - -
          Affymetrix Rat Gene 1.1 ST Array 10775896 - -
          Rat Genome 230 2.0 Array 1368760_PM_at - -
          Rat Genome 230 2.0 Array 1368760_at - -
          Affymetrix HT Rat Focus Array 1368760_at - -
          Affymetrix HT Rat Focus Array 1368760_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400011046.rn.1 TC1400001503.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400011047.rn.1 TC1400001503.rn.1 RTA TC
          Rat Expression Set 230 1368760_at - -
          Rat Expression Set 230 1368760_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775896 - -
          Rat Genome U34 Set U45965_at - -
          Rat Genome U34 Set U45965_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20807096 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352133 - -
          RatRef-12 Expression BeadChip ILMN_65741 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0