Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586466_m1
          See other DNM1L GE Assays ›
          Gene Symbol:
          Dnm1l
          Gene Name:
          dynamin 1-like
          Gene Aliases:
          DLP1, Dnml1, Drp1
          Chromosome Location:
          Chr.11: 88830968 - 88882138 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586466_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnm1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053655.3 NP_446107.2 9 - 10 1246 90
          XM_006248715.2 XP_006248777.1 10 - 11 1296 90
          XM_006248716.2 XP_006248778.1 10 - 11 1278 90
          XM_006248717.2 XP_006248779.1 10 - 11 1295 90
          XM_006248718.2 XP_006248780.1 9 - 10 1256 90
          XM_006248719.2 XP_006248781.1 9 - 10 1240 90
          XM_006248720.2 XP_006248782.1 10 - 11 1295 90
          XM_006248721.2 XP_006248783.1 10 - 11 1295 90
          XM_006248722.2 XP_006248784.1 9 - 10 1256 90
          XM_006248723.2 XP_006248785.1 10 - 11 1277 90
          XM_006248724.2 XP_006248786.1 9 - 10 1240 90
          XM_008768840.1 XP_008767062.1 10 - 11 1296 90
          XM_008768841.1 XP_008767063.1 10 - 11 1278 90
          XM_008768842.1 XP_008767064.1 9 - 10 1257 90
          XM_008768843.1 XP_008767065.1 10 - 11 1278 90
          XM_008768844.1 XP_008767066.1 9 - 10 1240 90
          XM_008768845.1 XP_008767067.1 10 - 11 1295 90
          XM_008768846.1 XP_008767068.1 9 - 10 1240 90
          XM_008768847.1 XP_008767069.1 10 - 11 1295 90
          GenBank mRNA AF132727.1 - 10 - 11 1168 90
          BC085843.1 - 9 - 10 1246 7135909 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001037.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnm1l

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          release of cytochrome c from mitochondria
          dynamin polymerization involved in mitochondrial fission
          endocytosis
          metabolic process
          regulation of mitochondrion organization
          peroxisome fission
          regulation of protein oligomerization
          positive regulation of apoptotic process
          mitochondrial fragmentation involved in apoptotic process
          positive regulation of protein secretion
          protein homotetramerization
          membrane fusion
          necroptotic process
          mitochondrion morphogenesis
          protein localization to mitochondrion
          positive regulation of mitochondrial fission
          mitochondrial membrane fission
          positive regulation of release of cytochrome c from mitochondria
          regulation of peroxisome organization
          regulation of mitophagy
          positive regulation of intrinsic apoptotic signaling pathway
          GTPase activity
          protein binding
          GTP binding
          lipid binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023704 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023704 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023704 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P330168 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P972824 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P381848 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P330170 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121215 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701441676 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90436 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51975 - -
          Agilent Rat cDNA Microarray (G4105A) 701441676 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755872 - -
          Affymetrix Rat Gene 1.0 ST Array 10755849 - -
          Affymetrix Rat Gene 1.0 ST Array 10755860 - -
          Affymetrix Rat Gene 1.0 ST Array 10755863 - -
          Affymetrix Rat Gene 1.0 ST Array 10755862 - -
          Affymetrix Rat Gene 1.0 ST Array 10755867 - -
          Affymetrix Rat Gene 1.0 ST Array 10755853 - -
          Affymetrix Rat Gene 1.0 ST Array 10755864 - -
          Affymetrix Rat Gene 1.0 ST Array 10755870 - -
          Affymetrix Rat Gene 1.0 ST Array 10755869 - -
          Affymetrix Rat Gene 1.0 ST Array 10755865 - -
          Affymetrix Rat Gene 1.0 ST Array 10755857 - -
          Affymetrix Rat Gene 1.0 ST Array 10755850 - -
          Affymetrix Rat Gene 1.0 ST Array 10755859 - -
          Affymetrix Rat Gene 1.0 ST Array 10755851 - -
          Affymetrix Rat Gene 1.0 ST Array 10755866 - -
          Affymetrix Rat Gene 1.0 ST Array 10755871 - -
          Affymetrix Rat Gene 1.0 ST Array 10755858 - -
          Affymetrix Rat Gene 1.0 ST Array 10755852 - -
          Affymetrix Rat Gene 1.0 ST Array 10755856 - -
          Affymetrix Rat Gene 1.0 ST Array 10755854 - -
          Affymetrix Rat Gene 1.0 ST Array 10755861 - -
          Affymetrix Rat Gene 1.1 ST Array 10755846 - -
          Rat Genome 230 2.0 Array 1383183_PM_at - -
          Rat Genome 230 2.0 Array 1394227_PM_at - -
          Rat Genome 230 2.0 Array 1386460_PM_x_at - -
          Rat Genome 230 2.0 Array 1388022_PM_a_at - -
          Rat Genome 230 2.0 Array 1383183_at - -
          Rat Genome 230 2.0 Array 1369220_at - -
          Rat Genome 230 2.0 Array 1386460_x_at - -
          Rat Genome 230 2.0 Array 1388022_a_at - -
          Rat Genome 230 2.0 Array 1369220_PM_at - -
          Rat Genome 230 2.0 Array 1394227_at - -
          Affymetrix HT Rat Focus Array 1369220_PM_at - -
          Affymetrix HT Rat Focus Array 1369220_at - -
          Affymetrix HT Rat Focus Array 1388022_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388022_a_at - -
          Affymetrix Rat Clariom S Assay PSR1100006255.rn.1 TC1100001037.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006256.rn.1 TC1100001037.rn.1 RTA TC
          Rat Expression Set 230 1369220_PM_at - -
          Rat Expression Set 230 1388022_PM_a_at - -
          Rat Expression Set 230 1369220_at - -
          Rat Expression Set 230 1388022_a_at - -
          Rat Expression Set 230 1386460_x_at - -
          Rat Expression Set 230 1383183_at - -
          Rat Expression Set 230 1383183_PM_at - -
          Rat Expression Set 230 1386460_PM_x_at - -
          Rat Expression Set 230 1394227_at - -
          Rat Expression Set 230 1394227_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755846 - -
          Rat Genome U34 Set AF020210_s_at - -
          Rat Genome U34 Set AF019043_at - -
          Rat Genome U34 Set AF020212_s_at - -
          Rat Genome U34 Set AF020211_s_at - -
          Rat Genome U34 Set rc_AI072458_at - -
          Rat Genome U34 Set rc_AI237251_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21043159 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365756 - -
          RatRef-12 Expression BeadChip ILMN_53626 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline