Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586593_m1
          See other CRCP GE Assays ›
          Gene Symbol:
          Crcp
          Gene Name:
          CGRP receptor component
          Gene Aliases:
          Cgrp-rcp
          Chromosome Location:
          Chr.12: 30125237 - 30160857 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586593_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crcp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053670.3 NP_446122.1 5 - 6 312 56
          GenBank mRNA AF440799.2 - 5 - 6 312 56
          BC059117.1 - 5 - 6 416 6891094 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001434.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crcp

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase III promoter
          transcription initiation from RNA polymerase III promoter
          neuropeptide signaling pathway
          aging
          innate immune response
          defense response to virus
          nucleotide binding
          RNA polymerase III activity
          calcitonin gene-related peptide receptor activity
          molecular_function
          DNA-directed RNA polymerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P546058 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P148280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P148280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P546058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P546058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P148280 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P546058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P148280 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P148280 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700948258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3082 - -
          Agilent Rat cDNA Microarray (G4105A) 700948258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761368 - -
          Affymetrix Rat Gene 1.0 ST Array 10761370 - -
          Affymetrix Rat Gene 1.0 ST Array 10761373 - -
          Affymetrix Rat Gene 1.0 ST Array 10761371 - -
          Affymetrix Rat Gene 1.0 ST Array 10761369 - -
          Affymetrix Rat Gene 1.0 ST Array 10761374 - -
          Affymetrix Rat Gene 1.0 ST Array 10761372 - -
          Affymetrix Rat Gene 1.1 ST Array 10761367 - -
          Rat Genome 230 2.0 Array 1369994_PM_at - -
          Rat Genome 230 2.0 Array 1369994_at - -
          Affymetrix HT Rat Focus Array 1369994_at - -
          Affymetrix HT Rat Focus Array 1369994_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200012135.rn.1 TC1200001434.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012132.rn.1 TC1200001434.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012133.rn.1 TC1200001434.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012134.rn.1 TC1200001434.rn.1 RTA TC
          Rat Expression Set 230 1369994_PM_at - -
          Rat Expression Set 230 1369994_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761367 - -
          Rat Genome U34 Set rc_AA799418_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22381268 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366139 - -
          RatRef-12 Expression BeadChip ILMN_56819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline