Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587172_m1
          See other UOX GE Assays ›
          Gene Symbol:
          Uox
          Gene Name:
          urate oxidase
          Gene Aliases:
          UOX-2, Uri, Uri2
          Chromosome Location:
          Chr.2: 252452282 - 252488497 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587172_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uox
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053768.2 NP_446220.1 4 - 5 486 81
          XM_006233450.2 XP_006233512.1 5 - 6 677 81
          GenBank mRNA BC088112.1 - 4 - 5 488 7325917 81
          FQ209540.1 - 4 - 5 499 81
          FQ209683.1 - 4 - 5 502 81
          FQ210147.1 - 3 - 4 408 81
          FQ210270.1 - 4 - 5 500 81
          FQ218233.1 - 4 - 5 502 81
          FQ218339.1 - 4 - 5 500 81
          FQ218518.1 - 4 - 5 500 81
          FQ218662.1 - 3 - 4 228 81
          FQ218801.1 - 4 - 5 500 81
          FQ219119.1 - 4 - 5 500 81
          FQ219150.1 - 4 - 5 500 81
          FQ219398.1 - 4 - 5 502 81
          FQ219405.1 - 4 - 5 502 81
          J03959.1 - 4 - 5 444 81
          M24396.1 - 4 - 5 487 81
          X13098.1 - 4 - 5 487 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002374.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uox

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleobase catabolic process
          urate catabolic process
          oxidation-reduction process
          urate oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P392482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P392482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P392482 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P392482 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P392482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819745 - -
          Affymetrix Rat Gene 1.0 ST Array 10819747 - -
          Affymetrix Rat Gene 1.0 ST Array 10819744 - -
          Affymetrix Rat Gene 1.0 ST Array 10819746 - -
          Affymetrix Rat Gene 1.0 ST Array 10819742 - -
          Affymetrix Rat Gene 1.0 ST Array 10819748 - -
          Affymetrix Rat Gene 1.0 ST Array 10819743 - -
          Affymetrix Rat Gene 1.0 ST Array 10819741 - -
          Affymetrix Rat Gene 1.1 ST Array 10819740 - -
          Rat Genome 230 2.0 Array 1387963_a_at - -
          Rat Genome 230 2.0 Array 1387963_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387963_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387963_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200016697.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016698.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016699.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016703.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016701.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016700.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016702.rn.1 TC0200002374.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016696.rn.1 TC0200002374.rn.1 RTA TC
          Rat Expression Set 230 1387963_PM_a_at - -
          Rat Expression Set 230 1387963_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819740 - -
          Rat Genome U34 Set J03959_at - -
          Rat Genome U34 Set J03959_g_at - -
          Rat Genome U34 Set X13098cds_s_at - -
          Rat Genome U34 Set rc_AA925223_r_at - -
          Rat Genome U34 Set rc_AI169989_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22225008 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363851 - -
          RatRef-12 Expression BeadChip ILMN_67021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline