Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587482_m1
          See other SNAP29 GE Assays ›
          Gene Symbol:
          Snap29
          Gene Name:
          synaptosomal-associated protein 29
          Gene Aliases:
          Gs32
          Chromosome Location:
          Chr.11: 87827633 - 87858107 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587482_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snap29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053810.3 NP_446262.3 2 - 3 511 88
          GenBank mRNA AF035822.1 - 2 - 3 521 88
          AF260577.1 - 2 - 3 434 88
          BC091693.1 - 2 - 3 511 7134635 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100002012.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snap29

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          synaptic vesicle priming
          synaptic vesicle fusion to presynaptic active zone membrane
          autophagosome maturation
          SNAP receptor activity
          protein binding
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P230030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800012 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19732 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P230030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P230030 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P230030 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426387 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P230030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28314 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755761 - -
          Affymetrix Rat Gene 1.0 ST Array 10755759 - -
          Affymetrix Rat Gene 1.0 ST Array 10755757 - -
          Affymetrix Rat Gene 1.0 ST Array 10755756 - -
          Affymetrix Rat Gene 1.0 ST Array 10755760 - -
          Affymetrix Rat Gene 1.0 ST Array 10755758 - -
          Affymetrix Rat Gene 1.0 ST Array 10755762 - -
          Affymetrix Rat Gene 1.1 ST Array 10755755 - -
          Rat Genome 230 2.0 Array 1373213_PM_at - -
          Rat Genome 230 2.0 Array 1369316_s_at - -
          Rat Genome 230 2.0 Array 1369316_PM_s_at - -
          Rat Genome 230 2.0 Array 1373213_at - -
          Affymetrix HT Rat Focus Array 1373213_PM_at - -
          Affymetrix HT Rat Focus Array 1373213_at - -
          Affymetrix HT Rat Focus Array 1369316_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369316_s_at - -
          Affymetrix Rat Clariom S Assay PSR1100013283.rn.1 TC1100002012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013284.rn.1 TC1100002012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013285.rn.1 TC1100002012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013281.rn.1 TC1100002012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013282.rn.1 TC1100002012.rn.1 RTA TC
          Rat Expression Set 230 1369316_PM_s_at - -
          Rat Expression Set 230 1373213_at - -
          Rat Expression Set 230 1373213_PM_at - -
          Rat Expression Set 230 1369316_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755755 - -
          Rat Genome U34 Set rc_AI010450_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21344066 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline