Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587821_m1
          See other SCG3 GE Assays ›
          Gene Symbol:
          Scg3
          Gene Name:
          secretogranin III
          Gene Aliases:
          SgIII
          Chromosome Location:
          Chr.8: 82582352 - 82641536 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587821_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scg3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053856.2 NP_446308.1 6 - 7 994 70
          GenBank mRNA U02983.1 - 6 - 7 996 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002948.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scg3

          Gene Ontology Categories:

          Process(es)

          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P339518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155415 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339518 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339518 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P339518 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12960 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P339518 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P559401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74182 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918742 - -
          Affymetrix Rat Gene 1.0 ST Array 10918745 - -
          Affymetrix Rat Gene 1.0 ST Array 10918740 - -
          Affymetrix Rat Gene 1.0 ST Array 10918746 - -
          Affymetrix Rat Gene 1.0 ST Array 10918750 - -
          Affymetrix Rat Gene 1.0 ST Array 10918748 - -
          Affymetrix Rat Gene 1.0 ST Array 10918743 - -
          Affymetrix Rat Gene 1.0 ST Array 10918739 - -
          Affymetrix Rat Gene 1.0 ST Array 10918744 - -
          Affymetrix Rat Gene 1.0 ST Array 10918747 - -
          Affymetrix Rat Gene 1.0 ST Array 10918741 - -
          Affymetrix Rat Gene 1.0 ST Array 10918749 - -
          Affymetrix Rat Gene 1.1 ST Array 10918738 - -
          Rat Genome 230 2.0 Array 1368656_at - -
          Rat Genome 230 2.0 Array 1368656_PM_at - -
          Affymetrix HT Rat Focus Array 1368656_at - -
          Affymetrix HT Rat Focus Array 1368656_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800023671.rn.1 TC0800002948.rn.1 RTA TC
          Rat Expression Set 230 1368656_at - -
          Rat Expression Set 230 1368656_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918738 - -
          Rat Genome U34 Set U02983_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22113783 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367903 - -
          RatRef-12 Expression BeadChip ILMN_56986 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline