Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588090_m1
          See other CGRRF1 GE Assays ›
          Gene Symbol:
          Cgrrf1
          Gene Name:
          cell growth regulator with ring finger domain 1
          Gene Aliases:
          Cgr19
          Chromosome Location:
          Chr.15: 23619176 - 23639622 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588090_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cgrrf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053899.1 NP_446351.1 2 - 3 342 62
          GenBank mRNA BC078763.1 - 2 - 3 358 7107668 62
          FQ214200.1 - 2 - 3 253 62
          FQ218613.1 - 2 - 3 357 62
          U66471.1 - 2 - 3 342 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000231.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cgrrf1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle arrest
          negative regulation of cell growth
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162879 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043431 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043431 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043431 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P611223 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700544391 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700296775 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19876 - -
          Agilent Rat cDNA Microarray (G4105A) 700544391 - -
          Agilent Rat cDNA Microarray (G4105A) 700296775 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779650 - -
          Affymetrix Rat Gene 1.0 ST Array 10779648 - -
          Affymetrix Rat Gene 1.0 ST Array 10779651 - -
          Affymetrix Rat Gene 1.0 ST Array 10779653 - -
          Affymetrix Rat Gene 1.0 ST Array 10779647 - -
          Affymetrix Rat Gene 1.0 ST Array 10779649 - -
          Affymetrix Rat Gene 1.0 ST Array 10779652 - -
          Affymetrix Rat Gene 1.1 ST Array 10779646 - -
          Rat Genome 230 2.0 Array 1387244_at - -
          Rat Genome 230 2.0 Array 1387244_PM_at - -
          Affymetrix HT Rat Focus Array 1387244_at - -
          Affymetrix HT Rat Focus Array 1387244_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500001605.rn.1 TC1500000231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001611.rn.1 TC1500000231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001610.rn.1 TC1500000231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001604.rn.1 TC1500000231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001609.rn.1 TC1500000231.rn.1 RTA TC
          Rat Expression Set 230 1387244_PM_at - -
          Rat Expression Set 230 1387244_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779646 - -
          Rat Genome U34 Set U66471_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21642820 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367213 - -
          RatRef-12 Expression BeadChip ILMN_61389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline