Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589194_m1
          See other EPN1 GE Assays ›
          Gene Symbol:
          Epn1
          Gene Name:
          Epsin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 72294153 - 72310217 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057136.1 NP_476477.1 3 - 4 687 110
          GenBank mRNA AF018261.1 - 3 - 4 687 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004777.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Epn1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          endocytosis
          Notch signaling pathway
          female pregnancy
          embryonic organ development
          transcription factor binding
          lipid binding
          ion channel binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156613 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13042 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13042 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19196 - -
          Agilent Rat cDNA Microarray (G4105A) 701567129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718796 - -
          Affymetrix Rat Gene 1.0 ST Array 10718797 - -
          Affymetrix Rat Gene 1.0 ST Array 10718802 - -
          Affymetrix Rat Gene 1.0 ST Array 10718793 - -
          Affymetrix Rat Gene 1.0 ST Array 10718799 - -
          Affymetrix Rat Gene 1.0 ST Array 10718801 - -
          Affymetrix Rat Gene 1.0 ST Array 10718800 - -
          Affymetrix Rat Gene 1.0 ST Array 10718794 - -
          Affymetrix Rat Gene 1.0 ST Array 10718792 - -
          Affymetrix Rat Gene 1.0 ST Array 10718798 - -
          Affymetrix Rat Gene 1.0 ST Array 10718795 - -
          Affymetrix Rat Gene 1.1 ST Array 10718791 - -
          Rat Genome 230 2.0 Array 1369453_at - -
          Rat Genome 230 2.0 Array 1369453_PM_at - -
          Affymetrix HT Rat Focus Array 1369453_at - -
          Affymetrix HT Rat Focus Array 1369453_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100037523.rn.1 TC0100004777.rn.1 RTA TC
          Rat Expression Set 230 1369453_at - -
          Rat Expression Set 230 1369453_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718791 - -
          Rat Genome U34 Set AF018261_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21770758 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351904 - -
          RatRef-12 Expression BeadChip ILMN_66806 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline