Search
Search
| Gene Symbol: Apoa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277264.1 | NP_001264193.1 | 2 - 3 | 168 | 96 | |
| NM_080576.2 | NP_542143.1 | 3 - 4 | 208 | 96 | ||
| XM_006242865.2 | XP_006242927.1 | 3 - 4 | 306 | 96 | ||
| GenBank mRNA | AF202887.1 | - | 2 - 3 | 153 | 96 | |
| AF202888.1 | - | 2 - 3 | 153 | 96 | ||
| BC089780.1 | - | 3 - 4 | 208 | 7371817 | 96 | |
| FQ210962.1 | - | 2 - 3 | 168 | 96 | ||
| FQ219463.1 | - | 2 - 3 | 168 | 96 | ||
Set Membership: |
|