Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590126_m1
          See other HNRNPH1 GE Assays ›
          Gene Symbol:
          Hnrnph1
          Gene Name:
          heterogeneous nuclear ribonucleoprotein H1
          Gene Aliases:
          Hnrph, Hnrph1
          Chromosome Location:
          Chr.10: 35872619 - 35879911 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590126_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnph1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080896.1 NP_543172.1 11 - 12 1190 112
          GenBank mRNA AF367468.1 - 11 - 12 1190 112
          BC099792.1 - 1239 112
          BC129087.1 - 1348 8363754 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000526.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnrnph1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          mRNA processing
          RNA splicing
          regulation of RNA splicing
          nucleotide binding
          nucleic acid binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038338 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042792 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P187144 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P500923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73510 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733245 - -
          Affymetrix Rat Gene 1.0 ST Array 10733252 - -
          Affymetrix Rat Gene 1.0 ST Array 10733251 - -
          Affymetrix Rat Gene 1.0 ST Array 10733255 - -
          Affymetrix Rat Gene 1.0 ST Array 10733257 - -
          Affymetrix Rat Gene 1.0 ST Array 10733250 - -
          Affymetrix Rat Gene 1.0 ST Array 10733253 - -
          Affymetrix Rat Gene 1.0 ST Array 10733247 - -
          Affymetrix Rat Gene 1.0 ST Array 10733248 - -
          Affymetrix Rat Gene 1.0 ST Array 10733254 - -
          Affymetrix Rat Gene 1.0 ST Array 10733256 - -
          Affymetrix Rat Gene 1.0 ST Array 10733244 - -
          Affymetrix Rat Gene 1.0 ST Array 10733249 - -
          Affymetrix Rat Gene 1.0 ST Array 10733246 - -
          Affymetrix Rat Gene 1.1 ST Array 10733243 - -
          Rat Genome 230 2.0 Array 1367666_PM_at - -
          Rat Genome 230 2.0 Array 1367666_at - -
          Affymetrix HT Rat Focus Array 1367666_at - -
          Affymetrix HT Rat Focus Array 1367666_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000004380.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004399.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004400.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004384.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004388.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004393.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004394.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004397.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004377.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004386.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004392.rn.1 TC1000000526.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004398.rn.1 TC1000000526.rn.1 RTA TC
          Rat Expression Set 230 1367666_at - -
          Rat Expression Set 230 1367666_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733243 - -
          Rat Genome U34 Set rc_AI103977_at - -
          Rat Genome U34 Set rc_AI136376_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21529625 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370138 - -
          RatRef-12 Expression BeadChip ILMN_55796 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline