Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592121_m1
          See other IL1RL2 GE Assays ›
          Gene Symbol:
          Il1rl2
          Gene Name:
          interleukin 1 receptor-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 47048915 - 47093679 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592121_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1rl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133575.1 NP_598259.1 9 - 10 1233 109
          GenBank mRNA U49066.1 - 9 - 10 1233 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000537.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1rl2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          positive regulation of interleukin-6 production
          positive regulation of T cell differentiation
          regulation of inflammatory response
          interleukin-1 receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134119 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134119 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134119 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P165908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922856 - -
          Affymetrix Rat Gene 1.0 ST Array 10922847 - -
          Affymetrix Rat Gene 1.0 ST Array 10922851 - -
          Affymetrix Rat Gene 1.0 ST Array 10922853 - -
          Affymetrix Rat Gene 1.0 ST Array 10922846 - -
          Affymetrix Rat Gene 1.0 ST Array 10922854 - -
          Affymetrix Rat Gene 1.0 ST Array 10922852 - -
          Affymetrix Rat Gene 1.0 ST Array 10922848 - -
          Affymetrix Rat Gene 1.0 ST Array 10922850 - -
          Affymetrix Rat Gene 1.0 ST Array 10922855 - -
          Affymetrix Rat Gene 1.0 ST Array 10922849 - -
          Affymetrix Rat Gene 1.1 ST Array 10922845 - -
          Rat Genome 230 2.0 Array 1387504_at - -
          Rat Genome 230 2.0 Array 1387504_PM_at - -
          Affymetrix HT Rat Focus Array 1387504_at - -
          Affymetrix HT Rat Focus Array 1387504_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900004387.rn.1 TC0900000537.rn.1 RTA TC
          Rat Expression Set 230 1387504_PM_at - -
          Rat Expression Set 230 1387504_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922845 - -
          Rat Genome U34 Set U49066_at - -
          Rat Genome U34 Set U49066_at - -
          Rat Genome U34 Set U49066_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21160491 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366873 - -
          RatRef-12 Expression BeadChip ILMN_62997 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline