Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592144_m1
          See other RAB26 GE Assays ›
          Gene Symbol:
          Rab26
          Gene Name:
          RAB26, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 13894271 - 13898578 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592144_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133580.1 NP_598264.1 1 - 2 134 71
          GenBank mRNA BC061984.1 - 2 - 3 442 5599180 71
          U18771.1 - 1 - 2 134 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002392.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab26

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle docking involved in exocytosis
          metabolic process
          protein secretion
          vesicle-mediated transport
          regulation of exocytosis
          Rab protein signal transduction
          exocrine system development
          Golgi to plasma membrane protein transport
          regulated exocytosis
          protein localization to plasma membrane
          GTPase activity
          GTP binding
          GMP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1022237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1022237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1022237 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1022237 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1022237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80441 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741135 - -
          Affymetrix Rat Gene 1.0 ST Array 10741141 - -
          Affymetrix Rat Gene 1.0 ST Array 10741139 - -
          Affymetrix Rat Gene 1.0 ST Array 10741142 - -
          Affymetrix Rat Gene 1.0 ST Array 10741134 - -
          Affymetrix Rat Gene 1.0 ST Array 10741138 - -
          Affymetrix Rat Gene 1.0 ST Array 10741137 - -
          Affymetrix Rat Gene 1.0 ST Array 10741140 - -
          Affymetrix Rat Gene 1.0 ST Array 10741136 - -
          Affymetrix Rat Gene 1.0 ST Array 10741133 - -
          Affymetrix Rat Gene 1.1 ST Array 10741112 - -
          Rat Genome 230 2.0 Array 1369004_at - -
          Rat Genome 230 2.0 Array 1369004_PM_at - -
          Affymetrix HT Rat Focus Array 1369004_at - -
          Affymetrix HT Rat Focus Array 1369004_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000021289.rn.1 TC1000002392.rn.1 RTA TC
          Rat Expression Set 230 1369004_PM_at - -
          Rat Expression Set 230 1369004_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741112 - -
          Rat Genome U34 Set U18771_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21268731 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374745 - -
          RatRef-12 Expression BeadChip ILMN_61577 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline