Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592734_m1
          See other SULF1 GE Assays ›
          Gene Symbol:
          Sulf1
          Gene Name:
          sulfatase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 5999520 - 6186901 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592734_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sulf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134378.2 NP_599205.1 4 - 5 586 117
          XM_008763486.1 XP_008761708.1 4 - 5 1158 117
          XM_008763487.1 XP_008761709.1 4 - 5 1312 117
          XM_008763488.1 XP_008761710.1 4 - 5 767 117
          GenBank mRNA AF230072.1 - 4 - 5 587 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002163.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sulf1

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          kidney development
          negative regulation of endothelial cell proliferation
          chondrocyte development
          glomerular filtration
          cell adhesion
          metabolic process
          positive regulation of vascular endothelial growth factor production
          esophagus smooth muscle contraction
          negative regulation of angiogenesis
          positive regulation of Wnt signaling pathway
          heparan sulfate proteoglycan metabolic process
          negative regulation of cell migration
          positive regulation of BMP signaling pathway
          glomerular basement membrane development
          glial cell-derived neurotrophic factor receptor signaling pathway
          limb joint morphogenesis
          regulation of fibroblast growth factor receptor signaling pathway
          negative regulation of fibroblast growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of smooth muscle cell proliferation
          embryonic skeletal system development
          cartilage development
          bone development
          innervation
          negative regulation of prostatic bud formation
          arylsulfatase activity
          calcium ion binding
          N-acetylglucosamine-6-sulfatase activity
          alkyl sulfatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P427346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75471 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P427346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P427346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P427346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P427346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92312 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89062 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35409 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875004 - -
          Affymetrix Rat Gene 1.0 ST Array 10874990 - -
          Affymetrix Rat Gene 1.0 ST Array 10874984 - -
          Affymetrix Rat Gene 1.0 ST Array 10874991 - -
          Affymetrix Rat Gene 1.0 ST Array 10874989 - -
          Affymetrix Rat Gene 1.0 ST Array 10874992 - -
          Affymetrix Rat Gene 1.0 ST Array 10874982 - -
          Affymetrix Rat Gene 1.0 ST Array 10874999 - -
          Affymetrix Rat Gene 1.0 ST Array 10874994 - -
          Affymetrix Rat Gene 1.0 ST Array 10875002 - -
          Affymetrix Rat Gene 1.0 ST Array 10874988 - -
          Affymetrix Rat Gene 1.0 ST Array 10875000 - -
          Affymetrix Rat Gene 1.0 ST Array 10874995 - -
          Affymetrix Rat Gene 1.0 ST Array 10874986 - -
          Affymetrix Rat Gene 1.0 ST Array 10874998 - -
          Affymetrix Rat Gene 1.0 ST Array 10875001 - -
          Affymetrix Rat Gene 1.0 ST Array 10874996 - -
          Affymetrix Rat Gene 1.0 ST Array 10875003 - -
          Affymetrix Rat Gene 1.0 ST Array 10874997 - -
          Affymetrix Rat Gene 1.0 ST Array 10874985 - -
          Affymetrix Rat Gene 1.0 ST Array 10874987 - -
          Affymetrix Rat Gene 1.0 ST Array 10874993 - -
          Affymetrix Rat Gene 1.0 ST Array 10874983 - -
          Affymetrix Rat Gene 1.1 ST Array 10874981 - -
          Rat Genome 230 2.0 Array 1373062_PM_at - -
          Rat Genome 230 2.0 Array 1368771_PM_at - -
          Rat Genome 230 2.0 Array 1368771_at - -
          Rat Genome 230 2.0 Array 1375492_at - -
          Rat Genome 230 2.0 Array 1375492_PM_at - -
          Rat Genome 230 2.0 Array 1373062_at - -
          Affymetrix HT Rat Focus Array 1375492_at - -
          Affymetrix HT Rat Focus Array 1368771_at - -
          Affymetrix HT Rat Focus Array 1375492_PM_at - -
          Affymetrix HT Rat Focus Array 1368771_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500016452.rn.1 TC0500002163.rn.1 RTA TC
          Rat Expression Set 230 1375492_PM_at - -
          Rat Expression Set 230 1368771_PM_at - -
          Rat Expression Set 230 1368771_at - -
          Rat Expression Set 230 1375492_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874981 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21581686 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348907 - -
          RatRef-12 Expression BeadChip ILMN_67009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline