Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593457_m1
          See other CDHR5 GE Assays ›
          Gene Symbol:
          Cdhr5
          Gene Name:
          cadherin-related family member 5
          Gene Aliases:
          Mucdhl, Mupcdh
          Chromosome Location:
          Chr.1: 214255118 - 214263849 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593457_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdhr5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138525.1 NP_612534.1 3 - 4 360 63
          XM_006230508.2 XP_006230570.1 3 - 4 688 63
          XM_006230509.2 XP_006230571.1 3 - 4 686 63
          GenBank mRNA AF221952.1 - 3 - 4 360 63
          BC098904.1 - 3 - 4 362 7444666 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006941.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdhr5

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding
          beta-catenin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P994385 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123314 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P994385 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13288 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13288 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P994385 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13288 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P994385 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P994385 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701822326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12980 - -
          Agilent Rat cDNA Microarray (G4105A) 701822326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726774 - -
          Affymetrix Rat Gene 1.0 ST Array 10726781 - -
          Affymetrix Rat Gene 1.0 ST Array 10726776 - -
          Affymetrix Rat Gene 1.0 ST Array 10726782 - -
          Affymetrix Rat Gene 1.0 ST Array 10726770 - -
          Affymetrix Rat Gene 1.0 ST Array 10726771 - -
          Affymetrix Rat Gene 1.0 ST Array 10726773 - -
          Affymetrix Rat Gene 1.0 ST Array 10726775 - -
          Affymetrix Rat Gene 1.0 ST Array 10726787 - -
          Affymetrix Rat Gene 1.0 ST Array 10726778 - -
          Affymetrix Rat Gene 1.0 ST Array 10726777 - -
          Affymetrix Rat Gene 1.0 ST Array 10726784 - -
          Affymetrix Rat Gene 1.0 ST Array 10726783 - -
          Affymetrix Rat Gene 1.0 ST Array 10726785 - -
          Affymetrix Rat Gene 1.0 ST Array 10726786 - -
          Affymetrix Rat Gene 1.0 ST Array 10726772 - -
          Affymetrix Rat Gene 1.0 ST Array 10726780 - -
          Affymetrix Rat Gene 1.0 ST Array 10726779 - -
          Affymetrix Rat Gene 1.1 ST Array 10726769 - -
          Rat Genome 230 2.0 Array 1387972_PM_at - -
          Rat Genome 230 2.0 Array 1387972_at - -
          Affymetrix HT Rat Focus Array 1387972_at - -
          Affymetrix HT Rat Focus Array 1387972_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100054740.rn.1 TC0100006941.rn.1 RTA TC
          Rat Expression Set 230 1387972_at - -
          Rat Expression Set 230 1387972_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726769 - -
          Rat Genome U34 Set rc_AI058395_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21147685 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355457 - -
          RatRef-12 Expression BeadChip ILMN_62290 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline