Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594189_m1
          See other GRASP GE Assays ›
          Gene Symbol:
          Grasp
          Gene Name:
          GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 142869785 - 142877551 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594189_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grasp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138894.1 NP_620249.1 1 - 2 305 113
          GenBank mRNA AF374272.1 - 1 - 2 305 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001866.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grasp

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          protein localization
          protein binding
          PDZ domain binding
          ADP-ribosylation factor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017319 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017319 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017319 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102292 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899383 - -
          Affymetrix Rat Gene 1.0 ST Array 10899382 - -
          Affymetrix Rat Gene 1.0 ST Array 10899379 - -
          Affymetrix Rat Gene 1.0 ST Array 10899385 - -
          Affymetrix Rat Gene 1.0 ST Array 10899380 - -
          Affymetrix Rat Gene 1.0 ST Array 10899386 - -
          Affymetrix Rat Gene 1.0 ST Array 10899384 - -
          Affymetrix Rat Gene 1.0 ST Array 10899381 - -
          Affymetrix Rat Gene 1.1 ST Array 10899378 - -
          Rat Genome 230 2.0 Array 1370723_PM_at - -
          Rat Genome 230 2.0 Array 1370723_at - -
          Affymetrix HT Rat Focus Array 1370723_PM_at - -
          Affymetrix HT Rat Focus Array 1370723_at - -
          Affymetrix Rat Clariom S Assay PSR0700014511.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014513.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014508.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014512.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014507.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014510.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014514.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014515.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014509.rn.1 TC0700001866.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014506.rn.1 TC0700001866.rn.1 RTA TC
          Rat Expression Set 230 1370723_at - -
          Rat Expression Set 230 1370723_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899378 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21415049 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371821 - -
          RatRef-12 Expression BeadChip ILMN_49985 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline