Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594376_m1
          See other DAD1 GE Assays ›
          Gene Symbol:
          Dad1
          Gene Name:
          defender against cell death 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 32868136 - 32887933 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594376_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dad1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138910.3 NP_620265.1 1 - 2 245 79
          GenBank mRNA BC061530.1 - 1 - 2 249 6917861 79
          FQ223332.1 - 1 - 2 254 79

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dad1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst development
          protein glycosylation
          apoptotic process
          response to nutrient
          response to drug
          negative regulation of apoptotic process
          molecular_function
          oligosaccharyl transferase activity
          dolichyl-diphosphooligosaccharide-protein glycotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P156317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P156317 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156317 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783532 - -
          Affymetrix Rat Gene 1.0 ST Array 10783530 - -
          Affymetrix Rat Gene 1.0 ST Array 10783531 - -
          Affymetrix Rat Gene 1.1 ST Array 10783529 - -
          Rat Genome 230 2.0 Array 1367465_at - -
          Rat Genome 230 2.0 Array 1367465_PM_at - -
          Affymetrix HT Rat Focus Array 1367465_PM_at - -
          Affymetrix HT Rat Focus Array 1367465_at - -
          Rat Expression Set 230 1367465_PM_at - -
          Rat Expression Set 230 1367465_at - -
          Rat Expression Set 230 1367465_at - -
          Rat Expression Set 230 1367465_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783529 - -
          Rat Genome U34 Set rc_AI013627_at - -
          Rat Genome U34 Set Y13336cds_g_at - -
          Rat Genome U34 Set Y13336cds_at - -
          Rat Genome U34 Set Y13336cds_at - -
          Rat Genome U34 Set Y13336cds_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21535995 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355566 - -
          RatRef-12 Expression BeadChip ILMN_65985 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline