Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595046_m1
          See other SIPA1L1 GE Assays ›
          Gene Symbol:
          Sipa1l1
          Gene Name:
          signal-induced proliferation-associated 1 like 1
          Gene Aliases:
          SPAL, Spa-1, Spa1, Spar
          Chromosome Location:
          Chr.6: 106052212 - 106163249 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595046_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sipa1l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139330.1 NP_647546.1 2844 123
          GenBank mRNA AF026504.1 - 2844 123
          AY043226.1 - 2407 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001124.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sipa1l1

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton reorganization
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          regulation of synaptic plasticity
          regulation of dendrite morphogenesis
          regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          regulation of dendritic spine morphogenesis
          GTPase activator activity
          protein binding
          protein kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          ephrin receptor binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152993 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152993 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152993 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045515 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702078310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68562 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97357 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35445 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19435 - -
          Agilent Rat cDNA Microarray (G4105A) 702078310 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885755 - -
          Affymetrix Rat Gene 1.0 ST Array 10885746 - -
          Affymetrix Rat Gene 1.0 ST Array 10885749 - -
          Affymetrix Rat Gene 1.0 ST Array 10885742 - -
          Affymetrix Rat Gene 1.0 ST Array 10885764 - -
          Affymetrix Rat Gene 1.0 ST Array 10885747 - -
          Affymetrix Rat Gene 1.0 ST Array 10885756 - -
          Affymetrix Rat Gene 1.0 ST Array 10885750 - -
          Affymetrix Rat Gene 1.0 ST Array 10885759 - -
          Affymetrix Rat Gene 1.0 ST Array 10885757 - -
          Affymetrix Rat Gene 1.0 ST Array 10885744 - -
          Affymetrix Rat Gene 1.0 ST Array 10885762 - -
          Affymetrix Rat Gene 1.0 ST Array 10885748 - -
          Affymetrix Rat Gene 1.0 ST Array 10885763 - -
          Affymetrix Rat Gene 1.0 ST Array 10885741 - -
          Affymetrix Rat Gene 1.0 ST Array 10885751 - -
          Affymetrix Rat Gene 1.0 ST Array 10885745 - -
          Affymetrix Rat Gene 1.0 ST Array 10885758 - -
          Affymetrix Rat Gene 1.0 ST Array 10885753 - -
          Affymetrix Rat Gene 1.0 ST Array 10885760 - -
          Affymetrix Rat Gene 1.0 ST Array 10885761 - -
          Affymetrix Rat Gene 1.0 ST Array 10885743 - -
          Affymetrix Rat Gene 1.0 ST Array 10885752 - -
          Affymetrix Rat Gene 1.1 ST Array 10885740 - -
          Rat Genome 230 2.0 Array 1370330_at - -
          Rat Genome 230 2.0 Array 1370330_PM_at - -
          Affymetrix HT Rat Focus Array 1370330_PM_at - -
          Affymetrix HT Rat Focus Array 1370330_at - -
          Affymetrix Rat Clariom S Assay PSR0600007806.rn.1 TC0600001124.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600007803.rn.1 TC0600001124.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600007804.rn.1 TC0600001124.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600007805.rn.1 TC0600001124.rn.1 RTA TC
          Rat Expression Set 230 1370330_PM_at - -
          Rat Expression Set 230 1370330_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885740 - -
          Rat Genome U34 Set AF026504_at - -
          Rat Genome U34 Set rc_AI237576_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22159927 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356407 - -
          RatRef-12 Expression BeadChip ILMN_57818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline