Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595142_m1
          See other SLC36A2 GE Assays ›
          Gene Symbol:
          Slc36a2
          Gene Name:
          solute carrier family 36 (proton/amino acid symporter), member 2
          Gene Aliases:
          Pat2, Tramd1
          Chromosome Location:
          Chr.10: 40497184 - 40525033 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595142_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc36a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139339.1 NP_647555.1 7 - 8 946 85
          GenBank mRNA AF512430.1 - 7 - 8 946 85
          BC166414.1 - 7 - 8 880 9027758 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc36a2

          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of proton transport
          L-alanine transport
          glycine transport
          proline transport
          proton transport
          amino acid export
          proline transmembrane transport
          regulation of proline transport
          positive regulation of glycine import
          hydrogen ion transmembrane transport
          hydrogen:amino acid symporter activity
          L-tyrosine transmembrane transporter activity
          hydrogen ion transmembrane transporter activity
          L-alanine transmembrane transporter activity
          glycine transmembrane transporter activity
          L-proline transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P337064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178875 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337064 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337064 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P337064 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742775 - -
          Affymetrix Rat Gene 1.0 ST Array 10742772 - -
          Affymetrix Rat Gene 1.0 ST Array 10742768 - -
          Affymetrix Rat Gene 1.0 ST Array 10742771 - -
          Affymetrix Rat Gene 1.0 ST Array 10742773 - -
          Affymetrix Rat Gene 1.0 ST Array 10742776 - -
          Affymetrix Rat Gene 1.0 ST Array 10742769 - -
          Affymetrix Rat Gene 1.0 ST Array 10742767 - -
          Affymetrix Rat Gene 1.0 ST Array 10742770 - -
          Affymetrix Rat Gene 1.0 ST Array 10742774 - -
          Affymetrix Rat Gene 1.1 ST Array 10742766 - -
          Rat Genome 230 2.0 Array 1393663_PM_at - -
          Rat Genome 230 2.0 Array 1393663_at - -
          Affymetrix HT Rat Focus Array 1393663_at - -
          Affymetrix HT Rat Focus Array 1393663_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000024794.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024796.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024799.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024801.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024792.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024798.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024795.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024797.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024803.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024793.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024800.rn.1 TC1000002823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000024802.rn.1 TC1000002823.rn.1 RTA TC
          Rat Expression Set 230 1393663_at - -
          Rat Expression Set 230 1393663_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742766 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21095217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline