Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595391_m1
          See other RETSAT GE Assays ›
          Gene Symbol:
          Retsat
          Gene Name:
          retinol saturase (all trans retinol 13,14 reductase)
          Gene Aliases:
          Rmt7
          Chromosome Location:
          Chr.4: 100465365 - 100474128 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Retsat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145084.1 NP_659552.1 5 - 6 997 79
          GenBank mRNA AF465614.1 - 5 - 6 997 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001126.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Retsat

          Gene Ontology Categories:

          Function(s) Process(es)

          retinol metabolic process
          oxidation-reduction process
          oxidoreductase activity
          all-trans-retinol 13,14-reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P402507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11895 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P402507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P402507 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P402507 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856429 - -
          Affymetrix Rat Gene 1.0 ST Array 10856430 - -
          Affymetrix Rat Gene 1.0 ST Array 10856435 - -
          Affymetrix Rat Gene 1.0 ST Array 10856434 - -
          Affymetrix Rat Gene 1.0 ST Array 10856426 - -
          Affymetrix Rat Gene 1.0 ST Array 10856425 - -
          Affymetrix Rat Gene 1.0 ST Array 10856428 - -
          Affymetrix Rat Gene 1.0 ST Array 10856431 - -
          Affymetrix Rat Gene 1.0 ST Array 10856427 - -
          Affymetrix Rat Gene 1.0 ST Array 10856432 - -
          Affymetrix Rat Gene 1.1 ST Array 10856424 - -
          Rat Genome 230 2.0 Array 1370806_PM_at - -
          Rat Genome 230 2.0 Array 1370806_at - -
          Affymetrix HT Rat Focus Array 1370806_PM_at - -
          Affymetrix HT Rat Focus Array 1370806_at - -
          Affymetrix Rat Clariom S Assay PSR0400008116.rn.1 TC0400001126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008117.rn.1 TC0400001126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008119.rn.1 TC0400001126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008118.rn.1 TC0400001126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008120.rn.1 TC0400001126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008115.rn.1 TC0400001126.rn.1 RTA TC
          Rat Expression Set 230 1370806_PM_at - -
          Rat Expression Set 230 1370806_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856424 - -
          Rat Genome U34 Set rc_AA955151_at - -
          Rat Genome U34 Set rc_AI171797_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22091811 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356474 - -
          RatRef-12 Expression BeadChip ILMN_50846 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline