Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596104_m1
          See other F7 GE Assays ›
          Gene Symbol:
          F7
          Gene Name:
          coagulation factor VII (serum prothrombin conversion accelerator)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 81824610 - 81834923 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596104_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152846.1 NP_690059.1 1 - 2 75 88
          GenBank mRNA AF532184.1 - 1 - 2 75 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002023.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F7

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          proteolysis
          blood coagulation
          circadian rhythm
          response to hormone
          positive regulation of platelet-derived growth factor receptor signaling pathway
          response to organic cyclic compound
          positive regulation of blood coagulation
          organ regeneration
          response to nutrient levels
          response to vitamin K
          response to estrogen
          positive regulation of positive chemotaxis
          positive regulation of protein kinase B signaling
          response to growth hormone
          glycoprotein binding
          endopeptidase activity
          serine-type endopeptidase activity
          receptor binding
          calcium ion binding
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P315947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105996 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P315947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P315947 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P315947 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P315947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792833 - -
          Affymetrix Rat Gene 1.0 ST Array 10792835 - -
          Affymetrix Rat Gene 1.0 ST Array 10792837 - -
          Affymetrix Rat Gene 1.0 ST Array 10792834 - -
          Affymetrix Rat Gene 1.0 ST Array 10792836 - -
          Affymetrix Rat Gene 1.0 ST Array 10792841 - -
          Affymetrix Rat Gene 1.0 ST Array 10792839 - -
          Affymetrix Rat Gene 1.0 ST Array 10792842 - -
          Affymetrix Rat Gene 1.0 ST Array 10792840 - -
          Affymetrix Rat Gene 1.0 ST Array 10792838 - -
          Affymetrix Rat Gene 1.1 ST Array 10792832 - -
          Affymetrix Rat Clariom S Assay PSR1600015018.rn.1 TC1600002023.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015020.rn.1 TC1600002023.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10792832 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184490 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358625 - -
          RatRef-12 Expression BeadChip ILMN_54569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline