Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596232_m1
          See other ALDH1A3 GE Assays ›
          Gene Symbol:
          Aldh1a3
          Gene Name:
          aldehyde dehydrogenase 1 family, member A3
          Gene Aliases:
          Aldh6
          Chromosome Location:
          Chr.1: 127302920 - 127337828 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596232_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh1a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153300.1 NP_695212.1 2 - 3 208 86
          GenBank mRNA AB369261.1 - 2 - 3 208 86
          AF434845.1 - 2 - 3 208 86
          BC166415.1 - 265 9025597 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005703.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh1a3

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          optic cup morphogenesis involved in camera-type eye development
          retinoic acid biosynthetic process
          locomotory behavior
          nucleus accumbens development
          pituitary gland development
          embryonic camera-type eye development
          inner ear morphogenesis
          response to drug
          retinol metabolic process
          retinoic acid metabolic process
          retinal metabolic process
          positive regulation of apoptotic process
          embryonic eye morphogenesis
          neuromuscular process controlling balance
          oxidation-reduction process
          righting reflex
          olfactory pit development
          face development
          retinal dehydrogenase activity
          aldehyde dehydrogenase (NAD) activity
          aldehyde dehydrogenase [NAD(P)+] activity
          protein homodimerization activity
          thyroid hormone binding
          NAD+ binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P273783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19871 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273783 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273783 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P273783 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P273783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57782 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69215 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10722667 - -
          Affymetrix Rat Gene 1.0 ST Array 10722659 - -
          Affymetrix Rat Gene 1.0 ST Array 10722666 - -
          Affymetrix Rat Gene 1.0 ST Array 10722660 - -
          Affymetrix Rat Gene 1.0 ST Array 10722662 - -
          Affymetrix Rat Gene 1.0 ST Array 10722657 - -
          Affymetrix Rat Gene 1.0 ST Array 10722664 - -
          Affymetrix Rat Gene 1.0 ST Array 10722671 - -
          Affymetrix Rat Gene 1.0 ST Array 10722665 - -
          Affymetrix Rat Gene 1.0 ST Array 10722661 - -
          Affymetrix Rat Gene 1.0 ST Array 10722656 - -
          Affymetrix Rat Gene 1.0 ST Array 10722658 - -
          Affymetrix Rat Gene 1.0 ST Array 10722669 - -
          Affymetrix Rat Gene 1.0 ST Array 10722668 - -
          Affymetrix Rat Gene 1.1 ST Array 10722655 - -
          Rat Genome 230 2.0 Array 1383469_at - -
          Rat Genome 230 2.0 Array 1383469_PM_at - -
          Affymetrix HT Rat Focus Array 1383469_PM_at - -
          Affymetrix HT Rat Focus Array 1383469_at - -
          Affymetrix Rat Clariom S Assay PSR0100045781.rn.1 TC0100005703.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045779.rn.1 TC0100005703.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045780.rn.1 TC0100005703.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045782.rn.1 TC0100005703.rn.1 RTA TC
          Rat Expression Set 230 1383469_at - -
          Rat Expression Set 230 1383469_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10722655 - -
          Rat Genome U34 Set rc_AA848809_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22091907 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352205 - -
          RatRef-12 Expression BeadChip ILMN_49413 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline